Skip to Content
Merck
All Photos(1)

Key Documents

798150

Sigma-Aldrich

Fluorescent nanodiamond

Nitrogen vacancy ~3ppm NV centers, 90 nm avg. part. size, 1 mg/mL in deionized water

Synonym(s):

Diamond nanopowder, Nanodiamonds

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
UNSPSC Code:
12352302
NACRES:
NA.23

form

suspension

Quality Level

composition

Nitrogen vacancy, ~3ppm NV centers

concentration

1 mg/mL in deionized water

avg. part. size

90 nm

fluorescence

λex 514 nm; λem 711 nm

Looking for similar products? Visit Product Comparison Guide

General description

Product can be functionalized with carboxylic acid, hydroxyl, ketones, ethers, and/or hydrogen.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Fluorescent Nanodiamonds Protein-Functionalized for Cell Labeling and Targeting
B-M Chang, H-S Lin, L-J Su, et al.
Advances in Functional Materials, 23(46), 5737-5745 (2013)
Nanodiamonds for optical bioimaging
Y. Y. Hui, C-L Cheng, and H-C Chang
Journal of Physics D: Applied Physics, 43(37), 374021-374021 (2010)
The Exocytosis of Fluorescent Nanodiamond and Its Use as a Long-Term Cell Tracker
Fang CY, et al.
Small, 7(23), 3363-3370 (2011)
Mass production and dynamic imaging of fluorescent nanodiamonds
Chang YR, et al.
Nature Nanotechnology, 3(5), 284-288 (2008)

Articles

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service