Skip to Content
Merck
All Photos(1)

Key Documents

79558

Sigma-Aldrich

Sodium phosphomolybdate hydrate

technical

Synonym(s):

Molybdophosphoric acid sodium salt, Phosphomolybdic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3[P(Mo3O10)4] · xH2O
CAS Number:
Molecular Weight:
1891.20 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

grade

technical

form

crystals

reaction suitability

core: molybdenum
reagent type: catalyst
reagent type: oxidant

mp

98 °C (dec.) (lit.)

density

2.83 g/mL at 25 °C (lit.)

SMILES string

O.[Na+].[Na+].[Na+].[O-][Mo](=O)(=O)O[Mo](=O)(=O)O[Mo](=O)(=O)O[Mo](=O)(=O)OP(=O)(O[Mo](=O)(=O)O[Mo](=O)(=O)O[Mo](=O)(=O)O[Mo]([O-])(=O)=O)O[Mo](=O)(=O)O[Mo](=O)(=O)O[Mo](=O)(=O)O[Mo]([O-])(=O)=O

InChI

1S/12Mo.3Na.H3O4P.H2O.36O/c;;;;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;;;;(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/q;;;;;;;;;6*+1;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;3*-1/p-3

InChI key

GVSXYXXCIUVHQT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • Supramolecular hydrogen-bonded Archimedean cages with encapsulated organic and inorganic species

Reactant for:
  • Temperature programmed reduction and hydrogen-isotope exchange
  • Passivation of stainless steel

Used for:
  • Plastic membrane electrodes for determination of flavoxate hydrochloride and cyclopentolate hydrochloride
  • Investigations of catalyst load role on transesterification of di-Me carbonate and phenol
  • Cocatalyst for methanol carbonylation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Almino Afonso de O Paiva et al.
International immunopharmacology, 11(9), 1241-1250 (2011-04-21)
This study analyzes the action of sulfated polysaccharides, fucans, from algae Lobophora variegata on zymosan-induced arthritis in rats. Groups of fucans, obtained after acetone fractionation (0.3-2.0 volumes), were denominated F0.3, F0.5, F0.8, F1, F1.5, and F2. The results that F1
Zhiming Cui et al.
Physical chemistry chemical physics : PCCP, 13(36), 16349-16357 (2011-08-16)
A simple self-assembly approach has been developed to functionalize carbon nanotubes (CNTs) with chitosan (CS) and heteropolyacids (HPAs) of phosphomolybdic acid (H(3)PMo(12)O(40), HPMo) and phosphotungstic acid (H(3)PW(12)O(40), HPW). The non-covalent functionalization method, which introduces homogenous surface functional groups with no
J K Ahn et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 69(9), 1294-1298 (2011-05-20)
We investigate the caesium concentrations in soils in mountain areas near Gori nuclear power plant in Korea, focusing on the measurement limits to the (134)Cs. In order to lower the minimum detectable amount (MDA) of activity for the (134)Cs, we
Zhiming Cui et al.
Physical chemistry chemical physics : PCCP, 14(37), 12823-12828 (2012-08-11)
It is challenging to simultaneously increase double layer- and pseudo-capacitance for supercapacitors. Phosphomolybdic acid/polyaniline/graphene nanocomposites (PMo(12)-PANI/GS) were prepared by using PMo(12) as a bifunctional reagent for not only well dispersing graphene for high electrochemical double layer capacitance but also in
Steven G Swarts et al.
Advances in experimental medicine and biology, 701, 291-297 (2011-03-30)
Fifty-eight semicarbazone and pyrazole derivatives of curcumin have been developed as potential mitigation agents to treat acute radiation syndrome (ARS). Pyridyl (D12, D13), furyl (D56), and phenyl (D68) derivatives of curcumin semi-carbazones were found to provide the highest dose modifying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service