Skip to Content
Merck
All Photos(2)

Key Documents

496626

Sigma-Aldrich

Sodium phosphotungstate hydrate

≥99.9% trace metals basis

Synonym(s):

Hydrated sodium phosphorus tungsten oxide, Sodium tungsten hydroxide oxide phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3PO4·12WO3·xH2O
CAS Number:
Molecular Weight:
2946.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
form:
powder

Quality Level

Assay

≥99.9% trace metals basis

form

powder

impurities

≤1000.0 ppm Trace Metal Analysis

mp

>350 °C (lit.)

SMILES string

O.[Na+].[Na+].[Na+].[O-]P([O-])(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W]([O-])(=O)=O

InChI

1S/3Na.H3O4P.H2O.36O.12W/c;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/q3*+1;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;-1;;;;;;;;;;;;+1/p-3

InChI key

ZJNHDCZHHPMFAZ-UHFFFAOYSA-K

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sigurd E Delanghe et al.
Journal of clinical medicine, 8(6) (2019-06-05)
The increased cardiovascular morbidity and mortality observed in chronic kidney disease (CKD) patients can be partly explained by the presence of carbamoylated lipoproteins. Lipid profiles can be determined with infrared spectroscopy. In this paper, the effects of carbamoylation on spectral
K Don D Gunaratne et al.
Physical chemistry chemical physics : PCCP, 18(13), 9021-9028 (2016-03-12)
Soft landing of mass-selected ions onto surfaces often results in partial loss of charge that may affect the structure and reactivity of deposited species. In this study, Keggin phosphotungstate anions in two selected charge states, PW12O40(3-) (WPOM(3-)) and PW12O40(2-) (WPOM(2-))
Vittoria Chimisso et al.
Soft matter, 17(3), 715-723 (2020-11-22)
Anionic poly(vinylcaprolactam-co-itaconicacid-co-dimethylitaconate) microgels were synthesized via dispersion polymerization and their responsiveness towards cations, namely Mg2+, Sr2+, Cu2+ and Fe3+, was investigated. The itaconic moieties chelate the metal ions which act as a crosslinker and decrease the electrostatic repulsion within the
Vittoria Chimisso et al.
Langmuir : the ACS journal of surfaces and colloids, 35(41), 13413-13420 (2019-10-05)
Poly(N-vinylcaprolactam-co-itaconate) (P(VCL-co-IADME) microgels were synthesized varying the molar ratio between VCL and IADME via free radical precipitation polymerization in the presence of quaternary ammonium surfactant. In order to determine the effect of the divalent metal ions on the structure and
Fu-Ying Shih et al.
Cancers, 12(2) (2020-02-27)
In this study, for the first time, we precisely assembled the poly-γ-benzyl-l-glutamate and an amphiphilic copolymer d-α-tocopherol polyethylene glycol succinate into a mixed micellar system for the embedment of the anticancer drug doxorubicin. Importantly, the intracellular drug-releasing behaviors could be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service