Skip to Content
Merck
All Photos(1)

Key Documents

793841

Sigma-Aldrich

2,2-Difluoroacetophenone

Synonym(s):

α,α-Difluoroacetophenone, 2,2-Difluoro-1-phenylethanone, Difluoromethyl phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6F2O
CAS Number:
Molecular Weight:
156.13
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

density

1.180±0.06 g/mL

functional group

fluoro
ketone
phenyl

SMILES string

FC(F)C(C1=CC=CC=C1)=O

InChI

1S/C8H6F2O/c9-8(10)7(11)6-4-2-1-3-5-6/h1-5,8H

InChI key

OLYKCPDTXVZOQF-UHFFFAOYSA-N

Application

Product was reported by Hartwig and coworkers in the one-pot synthesis of difluoromethylated arenes from aryl chlorides and bromides.


Pd-Catalyzed a-Arylation of a;a-Difluoroketones with Aryl Bromides and Chlorides. A Route to Difluoromethylarenes

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Professor Hartwig's research focuses on the discovery and understanding of new reactions catalyzed by transition metal complexes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service