Skip to Content
Merck
All Photos(1)

Key Documents

G2262

Sigma-Aldrich

γ-Glutamyltranspeptidase from porcine kidney

Type IV, 2.0-6.0 units/mg solid

Synonym(s):

γ-GT, γ-Glutamyltransferase, (5-Glutamyl)peptide:amino-acid 5-glutamyltransferase, GGTP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type IV

Quality Level

form

powder

specific activity

2.0-6.0 units/mg solid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

γ-Glutamyltranspeptidase is used in the food industry to improve the umami taste of soy sauce.

Biochem/physiol Actions

γ- Glutamyltranspeptidase cleaves theγ-glutamyl amide bond from glutathione and its S-conjugates to remove glutamate.

Quality

Crude

Unit Definition

One unit will liberate 1.0 μmole of p-nitroaniline from L-γ-glutamyl-p-nitroanilide per min at pH 8.5 at 25°C.

inhibitor

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving the umami taste of soy sauce by the addition of bacterial γ-glutamyltranspeptidase as a glutaminase to the fermentation mixture
K. Kijima & H. Suzuki
Enzyme and Microbial Technology, 41, 80-84 (2007)
S S Tate et al.
Molecular and cellular biochemistry, 39, 357-368 (1981-09-25)
gamma-Glutamyl transpeptidase catalyzes transfer of the gamma-glutamyl moiety of glutathione to amino acids, dipeptides, and to glutathione itself; the enzyme also catalyzes the hydrolysis of glutathione to glutamate and cysteinyl-glycine. This review deals with the tissue distribution and localization of
Mathias Oertli et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(8), 3047-3052 (2013-02-06)
Infection with the gastric bacterial pathogen Helicobacter pylori is typically contracted in early childhood and often persists for decades. The immunomodulatory properties of H. pylori that allow it to colonize humans persistently are believed to also account for H. pylori's
R Foddis et al.
Giornale italiano di medicina del lavoro ed ergonomia, 34(3 Suppl), 565-567 (2013-02-15)
The GGT enzyme, considered for years only as a marker of liver disease and alcohol abuse, has now revealed a risk of death for many causes. Through a molecular exclusion chromatography on FPLC system (Fast Protein Liquid Chromatography), it is
Stephanie Wickham et al.
The Biochemical journal, 450(3), 547-557 (2013-01-11)
GGT (γ-glutamyl transpeptidase) is an essential enzyme for maintaining cysteine homoeostasis, leukotriene synthesis, metabolism of glutathione conjugates and catabolism of extracellular glutathione. Overexpression of GGT has been implicated in many pathologies, and clinical inhibitors of GGT are under development for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service