Skip to Content
Merck
All Photos(1)

Key Documents

766917

Sigma-Aldrich

Lithium bis(trimethylsilyl)amide solution

1.5 M in THF

Synonym(s):

Hexamethyldisilazane lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NLi
CAS Number:
Molecular Weight:
167.33
Beilstein:
3567910
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.5 M in THF

density

0.893 g/mL at 25 °C

SMILES string

[Li]N([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C6H18NSi2.Li/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

YNESATAKKCNGOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium bis(trimethylsilyl)amide is commonly used in organic synthesis as a non-nucleophilic strong Bronsted base. It is soluble in most nonpolar solvents such as aromatic hydrocarbons, hexanes, and THF.

Application

Lithium bis(trimethylsilyl)amide can be used as a reagent:     
  • In the deprotonation and nucleophilic difluoromethylation reactions.
  • 3-methoxy substituted dihydropyrrole derivatives by reacting with aldehydes and lithiated methoxyallene via in situ formations of N-trimethylsilylated imines.  
  • In Darzens condensation and directed aldol condensation reactions.      
  • To synthesize poly(N-octyl-p-benzamide)s by chain-growth polycondensation of 4-octylaminobenzoic acid methyl ester.

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

31.3 °F - closed cup

Flash Point(C)

-0.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nur Hazlin Hazrin-Chong et al.
Journal of microbiological methods, 90(2), 96-99 (2012-05-09)
The use of hexamethyldisilazane (HMDS) as a drying agent was investigated in the specimen preparation for scanning electron microscopy (SEM) imaging of bacterial surface colonization on sub-bituminous coal. The ability of microbes to biofragment, ferment and generate methane from coal
Nan Li et al.
Lab on a chip, 8(12), 2105-2112 (2008-11-22)
High-density live cell array serves as a valuable tool for the development of high-throughput immunophenotyping systems and cell-based biosensors. In this paper, we have, for the first time, demonstrated a simple fabrication process to form the hexamethyldisilazane (HMDS) and poly(ethylene
Elsa Vennat et al.
Dental materials : official publication of the Academy of Dental Materials, 25(6), 729-735 (2009-01-29)
The objectives of this study were to assess demineralized dentin porosity and quantify the different porous features distribution within the material using mercury intrusion porosimetry (MIP) technique. We compared hexamethyldisilazane (HMDS) drying and lyophilization (LYO) (freeze-drying) in sample preparation. Fifty-six
Xiaoguang Li et al.
Chemical communications (Cambridge, England), 47(38), 10761-10763 (2011-08-30)
A silica microsphere suspension and a silica sol are employed in a two-step dipping process for the preparation of a superhydrophobic surface. It's not only a facile way to achieve the lotus effect, but can also create a multi-functional surface
Tatsuya Nitabaru et al.
Journal of the American Chemical Society, 131(38), 13860-13869 (2009-09-10)
Full details of an anti-selective catalytic asymmetric nitroaldol reaction promoted by a heterobimetallic catalyst comprised of Nd(5)O(O(i)Pr)(13), an amide-based ligand, and NaHMDS (sodium hexamethyldisilazide) are described. A systematic synthesis and evaluation of amide-based ligands led to the identification of optimum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service