Skip to Content
Merck
All Photos(2)

Key Documents

703516

Sigma-Aldrich

Zinc chloride solution

greener alternative

1.9 M in 2-methyltetrahydrofuran

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
liquid
Pricing and availability is not currently available.

form

liquid

Quality Level

reaction suitability

reagent type: catalyst
core: zinc

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1.9 M in 2-methyltetrahydrofuran
25 % (w/w)

density

1.07 g/mL at 25 °C

greener alternative category

SMILES string

Cl[Zn]Cl

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup - Solvent

Flash Point(C)

-12 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Molouk Beiromvand et al.
Preventive veterinary medicine, 109(1-2), 162-167 (2012-10-10)
Certain zoonotic parasites are enteropathogens in dogs that cause serious human disease such as cystic echinococcosis, human alveolar echinococcosis, visceral larva migrans, and ocular larva migrans. This study investigated the prevalence of intestinal parasites in dogs in the Chenaran County
Jessica Duprez et al.
PloS one, 7(10), e46831-e46831 (2012-10-12)
Rat pancreatic islet cell apoptosis is minimal after prolonged culture in 10 mmol/l glucose (G10), largely increased in 5 mmol/l glucose (G5) and moderately increased in 30 mmol/l glucose (G30). This glucose-dependent asymmetric V-shaped profile is preceded by parallel changes
Mayank Anand Gururani et al.
Plant science : an international journal of experimental plant biology, 198, 7-16 (2012-12-04)
Manganese stabilizing protein (MSP) is an important component of the Photosystem II (PSII) oxygen evolving complex. In our previous work, transgenic potato plants with reduced expression of MSP (MSP-As) were developed and their physiological and biochemical responses were studied. In
Russell E Carter et al.
Journal of neurochemistry, 125(5), 673-684 (2013-03-19)
Spreading depolarization (SD) is a feed-forward wave that propagates slowly throughout brain tissue and recovery from SD involves substantial metabolic demand. Presynaptic Zn(2+) release and intracellular accumulation occurs with SD, and elevated intracellular Zn(2+) ([Zn(2+) ]i ) can impair cellular
Elwin Molenbroek et al.
Dalton transactions (Cambridge, England : 2003), 42(8), 2973-2984 (2012-12-22)
In this work, the complexation of the bapbpy ligand to zinc dichloride is described (bapbpy = 6,6′-bis(2-aminopyridyl)-2,2′-bipyridine). The water-soluble, colorless complex [Zn(bapbpy)Cl]Cl·2H2O (compound 2·H2O) was synthesized; its X-ray crystal structure shows a mononuclear, pentacoordinated geometry with one chloride ligand in

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

  1. Can the Product No 703516 ZnCl2 in 1.9 M in 2-methyltetrahydrofuran be diluted with water or any buffer? Thank you- Demet

    1 answer
    1. Both zinc chloride and 2-methyltetrahydrofuran can be diluted in water.

      https://byjus.com/chemistry/zinc-chloride-zncl2/#:~:text=It%20is%20odourless.,acetone%2C%20ethanol%2C%20and%20glycerol.

      https://www.sigmaaldrich.com/sds/sial/673277?userType=anonymous

      Unfortunately, data characterizing the solubility of either compound in various buffers has not been determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service