Skip to Content
Merck
All Photos(1)

Key Documents

764493

Sigma-Aldrich

5-[4-(1,2,4,5-Tetrazin-3-yl)benzylamino]-5-oxopentanoic acid

97%

Synonym(s):

5-Oxo-5-[[[4-(1,2,4,5-tetrazin-3-yl)phenyl]methyl]amino]pentanoic acid, Benzylamino tetrazine acid, Bz-Tz-acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N5O3
CAS Number:
Molecular Weight:
301.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: click chemistry
reagent type: linker

mp

163-170 °C (decomposition)

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

OC(CCCC(NCC(C=C1)=CC=C1C2=NN=CN=N2)=O)=O

InChI

1S/C14H15N5O3/c20-12(2-1-3-13(21)22)15-8-10-4-6-11(7-5-10)14-18-16-9-17-19-14/h4-7,9H,1-3,8H2,(H,15,20)(H,21,22)

InChI key

KWNYIZNORNUUJQ-UHFFFAOYSA-N

Application

5-[4-(1,2,4,5-Tetrazin-3-yl)benzylamino]-5-oxopentanoic acid may be used in the synthesis of a PEG-tetrazine (PEG-Tz) macromer via acid-amine conjugation. This macromer can undergo click reaction with norbornene-functionalized peptides to form hydrogels that is useful for 3D cell culture.
Acid functionalized tetrazine for inverse electron demand Diels-Alder cycloaddition reactions. The tetrazine will react with strained alkenes such as transcyclooctene, norbornene and cyclopropene to yield a stable covalent linkage. Tetrazines have proven useful in bioorthogonal reactions for many biological imaging and bioconjugation applications.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile and efficient Lewis acid catalyzed synthesis of an asymmetric tetrazine useful for bio-orthogonal click chemistry applications
Alge DL, et al.
Tetrahedron Letters, 54(41), 5639-5641 (2013)
Synthetically tractable click hydrogels for three-dimensional cell culture formed using tetrazine?norbornene chemistry
Alge DL, et al.
Biomacromolecules, 14(4), 949-953 (2013)
18F-Labeling of Mannan for Inflammation Research with Positron Emission Tomography.
Li XG, et al.
ACS Medicinal Chemistry Letters, 7(9), 826-830 (2016)
Mark R Karver et al.
Bioconjugate chemistry, 22(11), 2263-2270 (2011-09-29)
1,2,4,5-Tetrazines have been established as effective dienes for inverse electron demand [4 + 2] Diels-Alder cycloaddition reactions with strained alkenes for over 50 years. Recently, this reaction pair combination has been applied to bioorthogonal labeling and cell detection applications; however
Neal K Devaraj et al.
Bioconjugate chemistry, 19(12), 2297-2299 (2008-12-05)
Bioorthogonal tetrazine cycloadditions have been applied to live cell labeling. Tetrazines react irreversibly with the strained dienophile norbornene forming dihydropyrazine products and dinitrogen. The reaction is high yielding, selective, and fast in aqueous media. Her2/neu receptors on live human breast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service