Skip to Content
Merck
All Photos(2)

Key Documents

900914

Sigma-Aldrich

Methyltetrazine-NHS ester

≥95%

Synonym(s):

2,5-Dioxo-1-pyrrolidinyl 4-(6-methyl-1,2,4,5-tetrazin-3-yl)benzeneacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N5O4
CAS Number:
Molecular Weight:
327.29
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

NHS ester

storage temp.

−20°C

SMILES string

O=C(ON1C(CCC1=O)=O)CC(C=C2)=CC=C2C3=NN=C(C)N=N3

InChI

1S/C15H13N5O4/c1-9-16-18-15(19-17-9)11-4-2-10(3-5-11)8-14(23)24-20-12(21)6-7-13(20)22/h2-5H,6-8H2,1H3

InChI key

BIHJLZOOHNOUCG-UHFFFAOYSA-N

Application

Methyltetrazine-NHS ester has an N-hydroxysuccinimide (NHS) ester that can be reacted with primary amines and a methyltetrazine, which is reactive to trans-cyclooctenes. The NHS ester reacts with primary amines (e.g. side chain of lysine residues or aminosilane-coated surfaces) at neutral or slightly basic pH to form covalent bonds. The short spacer arm adds minimal mass to modified molecules (230.2 daltons). It is generally used in click chemistry applications in bioconjugate chemistry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Throughput Chemical Probing of Full-Length Protein?Protein Interactions.
Song J M, et al.
ACS Combinatorial Science, 19(12), 763-769 (2017)
An anti-B cell maturation antigen bispecific antibody for multiple myeloma.
Ramadoss N S, et al.
Journal of the American Chemical Society, 137(16), 5288-5291 (2015)
A click chemistry-based microRNA maturation assay optimized for high-throughput screening.
Lorenz D A and Garner A L
Chemical Communications (Cambridge, England), 52(53), 8267-8270 (2016)
Biodegradable tetra-PEG hydrogels as carriers for a releasable drug delivery system.
Henise J, et al.
Bioconjugate Chemistry, 26(2), 270-278 (2015)

Articles

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Choosing the right crosslinker for your biochemical or bioconjugation research can be challenging. Our selection guide can help you find the perfect match for your applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service