Skip to Content
Merck
All Photos(1)

Key Documents

753688

Sigma-Aldrich

Gold nanoparticles

100 nm diameter, OD 1, stabilized suspension in 0.1 mM PBS, reactant free

Synonym(s):

Au NP, Gold colloid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanoparticles
suspension

concentration

~3.8E+9 particles/mL

OD

1

diameter

100 nm

λmax

565-574 nm

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanoparticles may be prepared by reduction of metal salt precursors with reducing agents, such as by citrate reduction of HAuCl4 in water.

Application

Gold nanoparticles are used in a wide range of applications. They may be used :
  • As conductors in printable inks,
  • In Photodynamic therapy,
  • In drug delivery and biomarkers,,
  • As calorimetric sensor,
  • In biological imaging,

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven D Perrault et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(25), 11194-11199 (2010-06-11)
Many small molecular anticancer agents are often ineffective at detecting or treating cancer due to their poor pharmacokinetics. Using nanoparticles as carriers can improve this because their large size reduces clearance and improves retention within tumors, but it also slows
Gold Nanoparticle Sensor for the Visual Detection of Pork Adulteration in Meatball Formulation
Ali ME, et al.
Journal of Nanomaterials (2012)
Tanya Stuchinskaya et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 10(5), 822-831 (2011-04-02)
A 4-component antibody-phthalocyanine-polyethylene glycol-gold nanoparticle conjugate is described for use as a potential drug for targeted photodynamic cancer therapy. Gold nanoparticles (4 nm) were stabilised with a self-assembled layer of a zinc-phthalocyanine derivative (photosensitiser) and a heterobifunctional polyethylene glycol. Anti-HER2
Plastic-Compatible Low Resistance Printable Gold Nanoparticle Conductors for Flexible Electronics
Huang D, et al.
Journal of the Electrochemical Society, 150, G412-G417 null
Gang Peng et al.
Nature nanotechnology, 4(10), 669-673 (2009-10-08)
Conventional diagnostic methods for lung cancer are unsuitable for widespread screening because they are expensive and occasionally miss tumours. Gas chromatography/mass spectrometry studies have shown that several volatile organic compounds, which normally appear at levels of 1-20 ppb in healthy

Articles

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

Sustainable energy sources with high production efficiency are crucial for meeting increasing energy demand.

Steven J. Oldenburg, Ph.D. provides an overview of lateral flow diagnostic assays and discusses the use of ultra-bright reporter particles based on the unique optical properties of gold nanoshells that significantly increase the sensitivity of lateral flow immunoassays.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service