Skip to Content
Merck
All Photos(2)

Key Documents

730734

Sigma-Aldrich

Trimethylgallium

packaged for use in deposition systems

Synonym(s):

Me3Ga, TMGa, TMG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ga(CH3)3
CAS Number:
Molecular Weight:
114.83
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

core: gallium

bp

92.5 °C/760 mmHg (lit.)

mp

-15.8 °C (lit.)

density

1.132 g/mL at 25 °C

SMILES string

C[Ga](C)C

InChI

1S/3CH3.Ga/h3*1H3;

InChI key

XCZXGTMEAKBVPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Atomic number of base material: 31 Gallium

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noémie Kraaijpoel et al.
PloS one, 15(2), e0228788-e0228788 (2020-02-12)
Compression ultrasonography (CUS) is the first-line imaging test in the diagnostic management of suspected deep vein thrombosis (DVT) of the lower extremity. Three CUS strategies are used in clinical practice. However, their relative diagnostic accuracy is uncertain. This systematic review
Mizuo Kajino et al.
GeoHealth, 4(9), e2020GH000259-e2020GH000259 (2020-10-02)
Emission inventories of anthropogenic transition metals, which contribute to aerosol oxidative potential (OP), in Asia (Δx = 0.25°, monthly, 2000-2008) and Japan (Δx = 2 km, hourly, mainly 2012) were developed, based on bottom-up inventories of particulate matters and metal profiles in a speciation database
Yuichi Manaka et al.
Scientific reports, 10(1), 2834-2834 (2020-02-20)
Ammonia from sewage and livestock manure is a major environmental pollutant. To consume environmental ammonia, we investigated the organic base-catalyzed synthesis of urea. 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) catalyzes the conversion of ammonium carbamate to urea in 35% yield at 100 °C. Moreover, DBU
C Guillén-Ponce et al.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico, 22(2), 201-212 (2020-01-26)
In the last 2 decades, clinical genetics on hereditary colorectal syndromes has shifted from just a molecular characterization of the different syndromes to the estimation of the individual risk of cancer and appropriate risk reduction strategies. In the last years, new
Adnan Al-Kaisy et al.
Trials, 21(1), 111-111 (2020-01-30)
Chronic neuropathic low back pain (CNLBP) is a debilitating condition in which established medical treatments seldom alleviate symptoms. Evidence demonstrates that high-frequency 10 kHz spinal cord stimulation (SCS) reduces pain and improves health-related quality of life in patients with failed back

Articles

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service