Skip to Content
Merck
All Photos(1)

Key Documents

752452

Sigma-Aldrich

Poly(ethylene glycol) diamine

average MN 3,000, cross-linking reagent carboxyl reactive

Synonym(s):

Poly(ethylene glycol) bis(amine), Polyethylene glycol, O,O′-Bis(2-aminoethyl)polyethylene glycol, Diaminopolyethylene glycol, PEG-diamine, Polyoxyethylene bis(amine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2CH2O)nCH2CH2NH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) diamine, average Mn 3,000

form

solid

mol wt

average Mn 3,000

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

mp

55-60 °C

polymer architecture

shape: linear
functionality: homobifunctional

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymer for preparing enzyme conjugates soluble in organic solvents†; promising drug carrier†

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malar A. Azagarsamy, et al.
Material Matters, 7 (2012)
C S Lee et al.
Artificial organs, 21(9), 1002-1006 (1997-09-01)
Various modifications of alginate-poly-L-lysine microcapsules were made, such as the inclusion of polyethylenimine (PEI) or carboxyl methyl cellulose (CMC) in the core and the coating of bis(polyoxyethylene bis[amine]) (PEGA) onto the microcapsule membrane surface. A characterization of the modified microcapsules
Alison G Patrick et al.
Macromolecular bioscience, 10(10), 1184-1193 (2010-07-02)
The design of hydrogels that simultaneously report protease activity and remove excess protease from solution is elucidated. The hydrogels, based on amino-PEGA, combine enzyme-specific peptides flanked with FRET complimented by charged amino acid residues that facilitate protease uptake via short
Gaëlle-Anne Cremer et al.
Journal of peptide science : an official publication of the European Peptide Society, 12(6), 437-442 (2006-01-25)
This paper describes the optimization of a synthesis of a difficult sequence related to a 12-mer sequence of a Pan DR epitope (PADRE). Elongation was followed by on-line monitoring of the N(alpha)-Fmoc removal adapted for the batch methodology. Studying the
Stephen J Connon et al.
Bioorganic & medicinal chemistry letters, 12(14), 1873-1876 (2002-06-28)
The synthesis and olefin metathesis activity in protic solvents of 7, a phosphine-free ruthenium alkylidene bound to a hydrophilic solid support are reported. This heterogeneous catalyst promotes relatively efficient ring closing- and cross-metathesis reactions in both methanol and water. The

Articles

Highlighting new synthetic modifications of PEG to improve the mechanical properties and degradation of resulting hydrogels in tissue engineering applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service