Skip to Content
Merck
All Photos(1)

Key Documents

731013

Sigma-Aldrich

JackiePhos

95%

Synonym(s):

2-{Bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy -2′,4′,6′-triisopropyl-1,1′-biphenyl, Bis(3,5-bis(trifluoromethyl)phenyl)(2′,4′,6′- triisopropyl-3,6-dimethoxybiphenyl-2-yl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H37F12O2P
CAS Number:
Molecular Weight:
796.66
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-X Bond Formation

mp

186-190 °C

functional group

phosphine

SMILES string

COc1ccc(OC)c(c1P(c2cc(cc(c2)C(F)(F)F)C(F)(F)F)c3cc(cc(c3)C(F)(F)F)C(F)(F)F)-c4c(cc(cc4C(C)C)C(C)C)C(C)C

InChI

1S/C39H37F12O2P/c1-19(2)22-11-29(20(3)4)33(30(12-22)21(5)6)34-31(52-7)9-10-32(53-8)35(34)54(27-15-23(36(40,41)42)13-24(16-27)37(43,44)45)28-17-25(38(46,47)48)14-26(18-28)39(49,50)51/h9-21H,1-8H3

InChI key

KYTUFIMHJNRPLC-UHFFFAOYSA-N

Application

Catalyst for N-Arylation reactions

Buchwald Phosphine Ligands for chemical Synthesis
JackiePhos is a Buchwald′s phosphine ligand which can be used for the:
  • glycosyl cross-coupling reaction of diaryliodonium triflates with anomeric stannanes to synthesize C-glycosides.
  • synthesis of gold-based catalyst complexes such as [Au(CH3CN)(JackiePhos)][SbF6] and [(JackiePhos)AuCl.
  • acylation of alkylcarbastannatranes.
  • synthesis of cyclic guanidines or cyclic ureas bearing dialkylaminomethyl groups in combination with Pd(acac)2 as a catalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gold-Catalyzed Intramolecular Regio-and Enantioselective Cycloisomerization of 1, 1-Bis (indolyl)-5-alkynes.
Huang L, et al.
Angewandte Chemie (International Edition in English), 125(26), 6899-6903 (2013)
Pd-Catalyzed Alkene Diamination Reactions of Nitrogen Electrophiles: Synthesis of Cyclic Guanidines and Ureas Bearing Dialkylaminomethyl Groups.
Peterson LJ, et al.
Organic Letters, 20(12), 3513-3517 (2018)
Gold-Catalyzed Cyclization of 1-(Indol-3-yl)-3-alkyn-1-ols: Facile Synthesis of Diversified Carbazoles.
Zhang Z, et al.
Chemistry?A European Journal , 19(32), 10625-10631 (2013)
Stereospecific Palladium-Catalyzed Acylation of Enantioenriched Alkylcarbastannatranes: A General Alternative to Asymmetric Enolate Reactions.
Wang CY, et al.
Angewandte Chemie (International Edition in English), 56(3), 856-860 (2017)
Glycosyl Cross-Coupling with Diaryliodonium Salts: Access to Aryl C-Glycosides of Biomedical Relevance.
Yi D, et al.
Organic Letters, 20(7), 1936-1940 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service