Skip to Content
Merck
All Photos(1)

Documents

791016

Sigma-Aldrich

RockPhos

97%

Synonym(s):

2-Di(tert-butyl)phosphino-2′,4′,6′-triisopropyl-3-methoxy-6-methylbiphenyl, Di-tert-butyl(2′,4′,6′-triisopropyl-3-methoxy-6-methyl-[1,1′-biphenyl]-2-yl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H49OP
CAS Number:
Molecular Weight:
468.69
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

121-131 °C

functional group

phosphine

storage temp.

15-25°C

SMILES string

CC1=C(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)C(P(C(C)(C)C)C(C)(C)C)=C(OC)C=C1

InChI

1S/C31H49OP/c1-19(2)23-17-24(20(3)4)28(25(18-23)21(5)6)27-22(7)15-16-26(32-14)29(27)33(30(8,9)10)31(11,12)13/h15-21H,1-14H3

InChI key

CVLLAKCGAFNZHJ-UHFFFAOYSA-N

Application

RockPhos is a biphenyl-based phosphine ligand used with Pd(0) catalyst for carbon-carbon and carbon-heteroatom bond formation reactions.
It can also be employed in the:
  • 2-fluroethoxylation of bromo-chalcones in the presence of palladium catalyst.
  • Conversion of aryl chlorides into aryl methoxides using RockPhos Pd G3 as a catalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Improved Synthesis of BrettPhos-and RockPhos-Type Biarylphosphine Ligands
Hoshiya N and Buchwald SL
advanced synthesis and catalysis, 354(10), 2031-2037 (2012)
P-chlorobenzyl ether: A p-methoxybenzyl ether in disguise
Viuff AH, et al.
Organic Letters, 18(22), 5773-5775 (2016)
A general and efficient Pd-catalyzed rapid 2-fluoroethoxylation of bromo-chalcones
Rangarajan TM, et al.
Journal of Fluorine Chemistry, 186(10), 101-110 (2016)
An Improved Synthesis of BrettPhos-and RockPhos-Type Biarylphosphine Ligands
Hoshiya N and Buchwald SL
Advanced Synthesis & Catalysis, 354(10), 2031-2037 (2012)

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service