713678
1,5-Cyclooctadiene{[dibenzyl((4S,5S)-5-methyl-2-phenyl-4,5-dihydro-4-oxazolyl)methyl]dicyclohexylphosphinite κN:κP}iridium(I) tetrakis(3,5-bis(trifluoromethyl)phenyl)borate
97%
Synonym(s):
Ubaphox, [((4S,5S)-Cy2-Ubaphox)Ir(COD)]BARF
About This Item
Recommended Products
Quality Level
Assay
97%
form
powder
optical purity
ee: ≥99%
SMILES string
[Ir+].[CH]1[CH]CC[CH][CH]CC1.C[C@@H]2OC(=N[C@@H]2C(Cc3ccccc3)(Cc4ccccc4)OP(C5CCCCC5)C6CCCCC6)c7ccccc7.FC(F)(F)c8cc(cc(c8)C(F)(F)F)[B-](c9cc(cc(c9)C(F)(F)F)C(F)(F)F)(c%10cc(cc(c%10)C(F)(F)F)C(F)(F)F)c%11cc(cc(c%11)C(F)(F)F)C(F)(F)F
InChI
1S/C37H46NO2P.C32H12BF24.C8H12.Ir/c1-29-35(38-36(39-29)32-21-11-4-12-22-32)37(27-30-17-7-2-8-18-30,28-31-19-9-3-10-20-31)40-41(33-23-13-5-14-24-33)34-25-15-6-16-26-34;34-25(35,36)13-1-14(26(37,38)39)6-21(5-13)33(22-7-15(27(40,41)42)2-16(8-22)28(43,44)45,23-9-17(29(46,47)48)3-18(10-23)30(49,50)51)24-11-19(31(52,53)54)4-20(12-24)32(55,56)57;1-2-4-6-8-7-5-3-1;/h2-4,7-12,17-22,29,33-35H,5-6,13-16,23-28H2,1H3;1-12H;1-2,7-8H,3-6H2;/q;-1;;+1/b;;2-1-,8-7-;/t29-,35-;;;/m0.../s1
InChI key
BGPHLJWQZLLYML-RDSUWHNOSA-N
General description
Application
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Andreas Pfaltz's chiral ligand designs, like semicorrins, pioneered bisoxazolines for widespread catalytic asymmetric synthesis.
Andreas Pfaltz's chiral ligand designs, like semicorrins, pioneered bisoxazolines for widespread catalytic asymmetric synthesis.
Andreas Pfaltz's chiral ligand designs, like semicorrins, pioneered bisoxazolines for widespread catalytic asymmetric synthesis.
Andreas Pfaltz's chiral ligand designs, like semicorrins, pioneered bisoxazolines for widespread catalytic asymmetric synthesis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service