Skip to Content
Merck
All Photos(1)

Key Documents

14694

Sigma-Aldrich

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Synonym(s):

Rhodium(I) tetrafluoroborate 1,5-Cyclooctadiene complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24BF4Rh
CAS Number:
Molecular Weight:
406.07
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

composition

carbon content, 45.7-49.0%

Quality Level

reaction suitability

core: rhodium
reagent type: catalyst

mp

~190 °C (dec.)

SMILES string

[Rh+].F[B-](F)(F)F.C1CC=CCCC=C1.C2CC=CCCC=C2

InChI

1S/2C8H12.BF4.Rh/c2*1-2-4-6-8-7-5-3-1;2-1(3,4)5;/h2*1-2,7-8H,3-6H2;;/q;;-1;+1/b2*2-1-,8-7-;;

InChI key

NBGSCPNNKGVTKU-QMDOQEJBSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Rhodium complex employed for preparing cationic COD-Rh complexes with phosphine ligands, e.g. for enantioselective hydrogenation; Hydrosilylation

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Takeuchi et al.
The Journal of Organic Chemistry, 60, 3045-3045 (1995)
U. Nagel et al.
Chemische Berichte, 119, 3326-3326 (1986)
I. Ojima et al.
Tetrahedron, 45, 6901-6901 (1989)
M. Sawamura et al.
Journal of the American Chemical Society, 117, 9602-9602 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service