Skip to Content
Merck
All Photos(1)

Documents

712728

Sigma-Aldrich

N-Tosylaziridine

98%

Synonym(s):

N-(p-Toluenesulfonyl)aziridine, N-(p-Tolylsulfonyl)aziridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2S
CAS Number:
Molecular Weight:
197.25
Beilstein:
154388
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)
98%

form

crystals

storage temp.

−20°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)N2CC2

InChI

1S/C9H11NO2S/c1-8-2-4-9(5-3-8)13(11,12)10-6-7-10/h2-5H,6-7H2,1H3

InChI key

VBNWSEVVMYMVLC-UHFFFAOYSA-N

Application

Reagent for beta-aminoethylation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anet, F.A.L.
Journal of the American Chemical Society, 89, 357-357 (1967)
Garcia-Espana, E.
Gazzetta Chimica Italiana, 115, 399-399 (1985)
Alexander M Gilchrist et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-12)
Synthetic anion transporters that facilitate chloride transport are promising candidates for channelopathy treatments. However, most anion transporters exhibit an undesired side effect of facilitating proton transport via interacting with fatty acids present in the membrane. To address the limitation, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service