Skip to Content
Merck

281069

Azetidine

98%

Synonym(s):

Trimethylene imine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H7N
CAS Number:
Molecular Weight:
57.09
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
207-963-8
Beilstein/REAXYS Number:
102384
MDL number:
Assay:
98%
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

98%

form

liquid

refractive index

n20/D 1.432 (lit.)

bp

61-62 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CNC1

InChI

1S/C3H7N/c1-2-4-3-1/h4H,1-3H2

InChI key

HONIICLYMWZJFZ-UHFFFAOYSA-N

General description

Phototransformations of azetidine radical cations in freonic matrices under the action of light with λ = 436nm has been investigated. The IR spectrum of azetidine in solid argon matrices has been measured.

Application

Azetidine was employed in:
  • a high yielding palladium-catalyzed cross-coupling raection with aryl bromides
  • Ullmann type coupling reaction with iodonitroflourenes


pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

-4.0 °F

flash_point_c

-20 °C

ppe

Faceshields, Gloves, Goggles



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Experimental and computational studies of the structure and vibrational spectra of azetidine derivatives.
Thompson CA, et al.
Journal of Molecular Structure, 491(1), 67-80 (1999)
Phototransformations of azetidine radical cations stabilized in freonic matrices.
Sorokin ID, et al.
High Energy Chemistry, 48(3), 180-184 (2014)
Synthesis, 243-243 (2007)



Global Trade Item Number

SKUGTIN
281069-250MG04061826214602
281069-1G04061838135223