Skip to Content
Merck
All Photos(1)

Key Documents

639095

Sigma-Aldrich

3-(Methylthio)propylamine

97%

Synonym(s):

1-Amino-3-(methylthio)propane, 3-(Methylmercapto)propylamine, 3-(Methylsulfanyl)propylamine, 3-(Methylthio)-1-propanamine, 3-(Methylthio)-1-propylamine, 3-Aminopropyl methyl sulfide, 3-Methylsulfanyl-1-propylamine, 3-Methylsulfanylpropan-1-amine, S-Methylhomocysteamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NS
CAS Number:
Molecular Weight:
105.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4940 (lit.)

bp

169 °C (lit.)

density

0.938 g/mL at 25 °C (lit.)

functional group

amine
thioether

SMILES string

CSCCCN

InChI

1S/C4H11NS/c1-6-4-2-3-5/h2-5H2,1H3

InChI key

KKYSBGWCYXYOHA-UHFFFAOYSA-N

Related Categories

Application

3-(Methylthio)propylamine can be used in the synthesis of rhodium(III)complexes involving acyclic diaminedithioether(DADTE)ligands.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"The complexation of rhodium (III) with acyclic diaminedithioether (DADTE) ligands"
Akgun Z, et al.
Dalton Transactions, 39(42), 10169-10178 (2010)
H F Emilie Heng et al.
Journal of food science and technology, 57(3), 1150-1157 (2020-03-04)
The aim of the study was to solve the mystery why sulfur-containing amino acids such as methionine can perform as an antioxidant during frying and hypothesized the antioxidative mechanisms. The results of this study revealed that sulfur-containing amino acids such
Ming-Fu Lin et al.
The Journal of chemical physics, 127(6), 064308-064308 (2007-08-21)
Photodissociation of 3-(methylthio)propylamine and cysteamine, the chromophores of S atom containing amino acid methionine and cysteine, respectively, was studied separately in a molecular beam at 193 nm using multimass ion imaging techniques. Four dissociation channels were observed for 3-(methylthio)propylamine, including
Olga B Morozova et al.
The journal of physical chemistry. A, 109(45), 10459-10466 (2006-07-13)
Using time-resolved chemically induced dynamic nuclear polarization (CIDNP) techniques, we have studied the mechanism of the photoreactions of triplet excited 4-carboxybenzophenone (CBP) with l-methionine (Met) and 3-(methylthio)propylamine (MTPA) in aqueous solution and the details of the formation of CIDNP at
H Hibasami et al.
Biochemical pharmacology, 38(21), 3673-3676 (1989-11-01)
Methylthiopropylamine (MTPA), an inhibitor of spermidine synthase, markedly induced ornithine decarboxylase (ODC) activity (about 30-fold of the basal level) in human lymphoid leukemia Molt 4B cells. This induction was blocked by the addition of spermidine, spermine or putrescine simultaneously with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service