61725
Vinyl laurate
≥99.0% (GC)
Synonym(s):
Dodecanoic acid vinyl ester, Vinyl dodecanoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)10COOCH=CH2
CAS Number:
Molecular Weight:
226.36
Beilstein:
1778369
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99.0% (GC)
refractive index
n20/D 1.441
bp
254 °C/1013 hPa (lit.)
density
0.871 g/mL at 20 °C (lit.)
functional group
ester
SMILES string
CCCCCCCCCCCC(=O)OC=C
InChI
1S/C14H26O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h4H,2-3,5-13H2,1H3
InChI key
GLVVKKSPKXTQRB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Vinyl laurate acts as an acyl donor during the transeterification of L-α-glycerophosphorylcholine (GPC) in the presence of Candida antarctica lipase B as catalyst to form lysophosphatidylcholine (LPC).
Application
Vinyl laurate can be used to synthesize the following:
- sucrose laurate esters from sucrose in organic solvents
- poly(vinyl laurate) (PVL)latexes
- porous conducting ‘‘soft′′ polymer foams which can be used as gas sensors
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
257.0 °F
Flash Point(C)
125 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lysophosphatidylcholine synthesis with Candida antarctica lipase B (Novozym 435)
Virto C and Adlercreutz P
Enzyme and Microbial Technology, 26(08), 630-635 (2000)
"Synthesis of sucrose laurate using a new alkaline protease"
Pedersen RN, et al.
Tetrahedron Asymmetry, 14(06), 667- 673 (2003)
Gabriele Spina et al.
IEEE journal of biomedical and health informatics, 19(5), 1567-1576 (2015-05-15)
With the growing amount of physical activity (PA) measures, the need for methods and algorithms that automatically analyze and interpret unannotated data increases. In this paper, PA is seen as a combination of multimodal constructs that can cooccur in different
Hugues Charest et al.
PloS one, 9(10), e109420-e109420 (2014-10-09)
HIV drug resistance represents a major threat for effective treatment. We assessed the trends in the frequency of drug resistance mutations and the monitored viral load (VL) in treatment-naïve (TN) and treatment-experienced (TE) individuals infected with HIV-1 in Québec, Canada
Sophie Cohen et al.
Journal of acquired immune deficiency syndromes (1999), 68(2), 178-185 (2014-11-19)
Immigrant HIV-infected adults in industrialized countries show a poorer clinical and virologic outcome compared with native patients. We aimed to investigate potential differences in clinical, immunological, and virologic outcome in Dutch HIV-infected children born in the Netherlands (NL) versus born
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service