Skip to Content
Merck
All Photos(1)

Key Documents

440213

Sigma-Aldrich

Triethoxy(octyl)silane

97%

Synonym(s):

Octyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7Si(OC2H5)3
CAS Number:
Molecular Weight:
276.49
Beilstein:
2325287
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

84-85 °C/0.5 mmHg (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC[Si](OCC)(OCC)OCC

InChI

1S/C14H32O3Si/c1-5-9-10-11-12-13-14-18(15-6-2,16-7-3)17-8-4/h5-14H2,1-4H3

InChI key

MSRJTTSHWYDFIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethoxy(octyl)silane (TEOS) is an alkoxide organosilane with four ethyl ester sidechains. It is used as a self-assembled monolayer and provides a hydrophobic coating with low surface energy. The water contact angle is in the range of 150-170°.

Application

Aluminium alloy can be coated with TEOS and 1H, 1H, 2H, 2H perfluorooctyl trichloro silane (PFOTS) to obtain superhydrophobicity with a high contact angle of 160-170° for vessel hulls and marine systems. It may be used to stabilize the porosity for a colorimetric sensor for immediate color changes, which finds potential application in the detection of toxic gases. It may also be used in the production of hybrid zeolites.
Triethoxy(octyl)silane may be used to prepare several formulations for fabricating low-cost, colorimetric sensor arrays to detect toxic gases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A colorimetric sensor array of porous pigments.
Lim H, et al.
Analyst, 134(12), 2453-2457 (2009)
Sun Li Chong et al.
Bioresource technology, 293, 122009-122009 (2019-09-08)
Here, we demonstrated the immobilization of bacterial feruloyl esterase (FAE) from Butyrivibrio sp. XPD2006, Lactobacillus crispatus, Butyrivibrio sp. AE2015, Ruminococcus albus, Cellulosilyticum ruminicola and Clostridium cellulovorans on SBA-15 and their ability to synthesize butyl ferulate (BFA). The BFae2 from Butyrivibrio
Fabrication of Superhydrophobic Al5083 Aluminum Alloy for Marine Applications.
Javad F, et al.
Protection of Metals and Physical Chemistry of Surfaces, 54(5), 899-908 (2018)
Danielle R Scheff et al.
Soft matter, 16(24), 5659-5668 (2020-06-11)
Macromolecules can phase separate to form liquid condensates, which are emerging as critical compartments in fields as diverse as intracellular organization and soft materials design. A myriad of macromolecules, including the protein FUS, form condensates which behave as isotropic liquids.
Synthesis of hybrid zeolites using a solvent-free method in the presence of different organosilanes.
Petkowicz I, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 241(12), 98-106 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service