Skip to Content
Merck
All Photos(1)

Documents

55665

Sigma-Aldrich

3,4-Dihydro-2H-pyran-2-methanol

≥98.5% (GC)

Synonym(s):

2-Hydroxymethyl-3,4-dihydro-2H-pyran, 3,4-Dihydro-2H-pyran-2-ylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein:
107275
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.5% (GC)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.478 (lit.)
n20/D 1.479

bp

92-93 °C/25 mmHg (lit.)

density

1.101 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC1CCC=CO1

InChI

1S/C6H10O2/c7-5-6-3-1-2-4-8-6/h2,4,6-7H,1,3,5H2

InChI key

XMICBFRKICBBKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Preparation of supported dihydropyran for mild immobilization and cleavage of alcohols

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preparation of supported dihydropyran for mild immobilization and cleavage of alcohols.
L.A. Thompson et al.
Tetrahedron Letters, 35, 9333-9333 (1994)
Facile Preparation of 2,6-Disubstituted Purines Using Solid-Phase Chemistry.
David A. Nugiel et al.
The Journal of organic chemistry, 62(1), 201-203 (1997-01-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service