Skip to Content
Merck
All Photos(2)

Key Documents

464058

Sigma-Aldrich

D-Glucal

96%

Synonym(s):

1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

optical activity

[α]21/D −7°, c = 1.9 in H2O

impurities

<1% methyl alcohol

mp

58-60 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@H]1OC=C[C@@H](O)[C@@H]1O

InChI

1S/C6H10O4/c7-3-5-6(9)4(8)1-2-10-5/h1-2,4-9H,3H2/t4-,5-,6+/m1/s1

InChI key

YVECGMZCTULTIS-PBXRRBTRSA-N

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi-Kai Cui et al.
Carbohydrate research, 358, 19-22 (2012-08-03)
2-Deoxyglycosides were synthesized in high α-selectivity by the direct addition of alcohols to D-glucal and D-galactal catalyzed by TMSI and PPh(3). The acid labile isopropylidene group is tolerated under this condition.
Syed Khalid Yousuf et al.
The Journal of organic chemistry, 75(9), 3097-3100 (2010-04-13)
Novel one-pot three- and four-component transformations of D-glucal to furan-based hydroxy triazole glycoconjugates have been achieved by sequential addition of reagents in the presence of Cu(OTf)(2)-Cu powder as catalysts. In general the carbohydrate-derived products were formed with high diastereomeric purity.
A possible catalytic role of pyridoxal-5'-phosphate in a general acid base catalyzed reaction of D-glucal with glycogen phosphorylases.
E J Helmreich et al.
Progress in clinical and biological research, 102 Pt C, 23-35 (1982-01-01)
Patricia Wildberger et al.
Carbohydrate research, 356, 224-232 (2012-05-18)
Cellobiose phosphorylase from Cellulomonas uda (CuCPase) is shown to utilize D-glucal as slow alternative donor substrate for stereospecific glycosyl transfer to inorganic phosphate, giving 2-deoxy-α-D-glucose 1-phosphate as the product. When performed in D(2)O, enzymatic phosphorolysis of D-glucal proceeds with incorporation
H W Klein et al.
Biochemistry, 21(26), 6675-6684 (1982-12-21)
D-Glucal, containing a highly reactive double bond, can replace glucose 1-phosphate as the glucosyl donor in phosphorylase-catalyzed glucosyl transfer to a suitable oligo- or polysaccharide acceptor: D-glucal + Pi + (glucose)Pi leads to n 2-deoxy-alpha-D-glucosyl(glucose)n in equilibrium 2-deoxy-alpha-D-glucose-1-P + (glucose)n.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service