Skip to Content
Merck
All Photos(1)

Key Documents

553913

Sigma-Aldrich

Magnesium boride

powder, −100 mesh, ≥99% trace metals basis

Synonym(s):

Magnesium diboride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgB2
CAS Number:
Molecular Weight:
45.93
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99% trace metals basis

form

powder

particle size

−100 mesh

SMILES string

[Mg]1B=B1

InChI

1S/B2.Mg/c1-2;/q+2;-2

InChI key

DRCUCSCGQLIENV-UHFFFAOYSA-N

General description

Magnesium boride (MgB2) powder is a superconductive material which has a critical temperature (Tc) = 39K. It can be used for a variety of applications such as hydrogen storage, high energy fuel and high field superconductors.

Application


  • Combustion characteristics of magnesium borides and their agglomerated particles: This research examines the combustion properties of magnesium borides, comparing them to boron to understand their potential in energetic materials (Y Guo, X Zhou, W Zhang, L Deng, Y Du, S Cheng, 2019).

  • Review on magnesium diboride (MgB2) as excellent superconductor: Discusses the superconducting properties of magnesium diboride and the effects of different production techniques on these properties, critical for applications in superconducting technologies (M Rafieazad, O Balci, S Acar, M Somer, 2017).

  • Novel magnesium borides and their superconductivity: Investigates new forms of magnesium borides under high pressure, exploring their potential superconducting properties, which could lead to new advancements in materials science (MMD Esfahani, Q Zhu, H Dong, AR Oganov, 2017).

Features and Benefits

Superconduction material with unique properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mingjian Fan et al.
Advanced healthcare materials, 8(13), e1900157-e1900157 (2019-04-11)
The hydrogen molecule is recognized as a high potential to attenuate toxic side effects of chemotherapy and also enhance chemotherapeutic efficacy, and the development of a novel hydrogen-generating prodrug for facile, safe, and efficient hydrogen delivery is vitally important for
D Eyidi et al.
Micron (Oxford, England : 1993), 34(2), 85-96 (2003-06-13)
Superconducting MgB(2) ceramics were prepared and yield superconducting transition temperatures of about 39 K. For covering the various length scales on which inhomogeneities appear in MgB(2), electron-probe micro-analysis (EPMA) and analytical transmission electron microscopy (TEM) were applied for a phase
Tailored materials for high-performance MgB(2) wire.
Jung Ho Kim et al.
Advanced materials (Deerfield Beach, Fla.), 23(42), 4942-4946 (2011-10-01)
B A Katsnel'son et al.
Gigiena truda i professional'nye zabolevaniia, (5)(5), 39-42 (1992-01-01)
Multichamber toxicokinetic model studies of the resorption, distribution and excretion properties of poorly soluble substances in subchronic action of suspensions administered intragastrically can be the first step in prognosing the toxicokinetic properties of the inhaled aerosols of the substances mostly
Superconductivity at 39 K in magnesium diboride
Nagamatsu J, et al.
Nature, 410(6824), 63-63 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service