Skip to Content
Merck
All Photos(1)

Key Documents

540099

Sigma-Aldrich

(R)-(+)-Styrene oxide

97%, optical purity ee: 97% (GLC)

Synonym(s):

(R)-(+)-Phenyloxirane, (R)-Phenylethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
Beilstein:
1984
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +33°, neat

optical purity

ee: 97% (GLC)

expl. lim.

~22 %

refractive index

n20/D 1.534 (lit.)

bp

89-90 °C/23 mmHg (lit.)

density

1.051 g/mL at 25 °C (lit.)

functional group

ether
phenyl

SMILES string

C1O[C@@H]1c2ccccc2

InChI

1S/C8H8O/c1-2-4-7(5-3-1)8-6-9-8/h1-5,8H,6H2/t8-/m0/s1

InChI key

AWMVMTVKBNGEAK-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(+)-Styrene oxide can be used:
  • As a chiral initiator in the synthesis of 5-pyrimidyl alkanol from its corresponding aldehyde using diisopropylzinc.
  • As a substrate in the stereoselective intermolecular O-alkylation of phenols with epoxides via Friedel–Crafts-type reaction.
  • As a substrate in the ring-opening reactions of epoxides with alcohols, carboxylic acids, and thiols using the AlPW12O40 catalyst.

Used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine.

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Carc. 1B - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new regio-and stereoselective intermolecular Friedel-Crafts alkylation of phenolic substrates with aryl epoxides
Bertolini F, et al.
Tetrahedron Letters, 47(1), 61-64 (2006)
Kang, B.; Chang, S.
Tetrahedron, 60, 7353-7353 (2004)
Aluminumdodecatungstophosphate (AlPW12O40) as a reusable Lewis acid catalyst: Facile regioselective ring opening of epoxides with alcohols, acetic acid and thiols
Firouzabadi H, et al.
J. Mol. Catal. A: Chem., 250(1-2), 237-242 (2006)
Enantioselective synthesis induced by chiral epoxides in conjunction with asymmetric autocatalysis
Kawasaki T, et al.
Tetrahedron Asymmetry, 15(23), 3699-3701 (2004)
Gary P Carlson
Drug and chemical toxicology, 34(4), 440-444 (2011-07-14)
Styrene is known to be hepatotoxic and pneumotoxic in rodents, and these adverse effects are related to its metabolism. Mice deficient in the enzymes responsible for both the activation and detoxification of styrene are useful in examining this relationship more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service