184802
4-Nitrophenethylamine hydrochloride
95%
Synonym(s):
2-(4-Nitrophenyl)ethylamine hydrochloride
About This Item
Recommended Products
Quality Level
Assay
95%
form
solid
mp
200 °C (dec.) (lit.)
functional group
amine
nitro
SMILES string
Cl.NCCc1ccc(cc1)[N+]([O-])=O
InChI
1S/C8H10N2O2.ClH/c9-6-5-7-1-3-8(4-2-7)10(11)12;/h1-4H,5-6,9H2;1H
InChI key
JVMHULJEYUQYSH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- ortho-metalated primary phenethylamines complexes containing six-membered palladacycles
- N-(4-nitrophenethyl)formamide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service