D128406
Didodecyl 3,3′-thiodipropionate
>97%
Synonym(s):
Thiodipropionic acid dilauryl ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
S[CH2CH2CO2(CH2)11CH3]2
CAS Number:
Molecular Weight:
514.84
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
0.2 mmHg ( 163 °C)
Assay
>97%
form
powder
mp
40-42 °C (lit.)
density
0.915 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCC
InChI
1S/C30H58O4S/c1-3-5-7-9-11-13-15-17-19-21-25-33-29(31)23-27-35-28-24-30(32)34-26-22-20-18-16-14-12-10-8-6-4-2/h3-28H2,1-2H3
InChI key
GHKOFFNLGXMVNJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
E A Eklund et al.
The Journal of biological chemistry, 265(15), 8426-8430 (1990-05-25)
Intact neutrophils possess a cellular mechanism that efficiently deactivates the microbicidal O2-generating NADPH oxidase during the respiratory burst (Akard, L. P., English, D., and Gabig, T. G. (1988) Blood 72, 322-327). The present studies directed at identifying the molecular mechanism(s)
Charlène Pouech et al.
Journal of chromatography. A, 1423, 111-123 (2015-11-12)
Polymer items are extensively present in the human environment. Humans may be consequently exposed to some compounds, such as additives, incorporated in these items. The objective of this work is to assess the human exposure to the main additives such
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service