Skip to Content
Merck
All Photos(1)

Documents

518174

Sigma-Aldrich

Tin(II) oxide

99.99% trace metals basis

Synonym(s):

Stannous oxide, Tin monoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnO
CAS Number:
Molecular Weight:
134.71
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

application(s)

battery manufacturing

SMILES string

O=[SnH2]

InChI

1S/O.Sn

InChI key

QHGNHLZPVBIIPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroki Tanaka et al.
PloS one, 10(8), e0134386-e0134386 (2015-08-27)
The aortic wall is perfused by the adventitial vasa vasorum (VV). Tissue hypoxia has previously been observed as a manifestation of enlarged abdominal aortic aneurysms (AAAs). We sought to determine whether hypoperfusion of the adventitial VV could develop AAAs. We
Andreas Römpp et al.
Analytical and bioanalytical chemistry, 407(8), 2329-2335 (2015-01-13)
Mass spectrometry imaging has become a popular tool for probing the chemical complexity of biological surfaces. This led to the development of a wide range of instrumentation and preparation protocols. It is thus desirable to evaluate and compare the data
Sean P Hill et al.
The journal of physical chemistry letters, 6(22), 4510-4517 (2015-11-05)
Molecular photon upconversion via triplet-triplet annihilation (TTA-UC), combining two or more low energy photons to generate a higher energy excited state, is an intriguing strategy to surpass the maximum efficiency for a single junction solar cell (<34%). Here, we introduce
Jixian Xu et al.
Nature communications, 6, 7081-7081 (2015-05-09)
Solution-processed planar perovskite devices are highly desirable in a wide variety of optoelectronic applications; however, they are prone to hysteresis and current instabilities. Here we report the first perovskite-PCBM hybrid solid with significantly reduced hysteresis and recombination loss achieved in
Ji-Wook Jang et al.
Nature communications, 6, 7447-7447 (2015-06-17)
Photoelectrochemical (PEC) water splitting promises a solution to the problem of large-scale solar energy storage. However, its development has been impeded by the poor performance of photoanodes, particularly in their capability for photovoltage generation. Many examples employing photovoltaic modules to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service