Merck
  • Photon upconversion and photocurrent generation via self-assembly at organic-inorganic interfaces.
Photon upconversion and photocurrent generation via self-assembly at organic-inorganic interfaces.
The journal of physical chemistry letters (2015-11-05)
Sean P Hill, Tanmay Banerjee, Tristan Dilbeck, Kenneth Hanson
ABSTRACT
Molecular photon upconversion via triplet-triplet annihilation (TTA-UC), combining two or more low energy photons to generate a higher energy excited state, is an intriguing strategy to surpass the maximum efficiency for a single junction solar cell (<34%). Here, we introduce self-assembled bilayers on metal oxide surfaces as a strategy to facilitate TTA-UC emission and demonstrate direct charge separation of the upconverted state. A 3-fold enhancement in transient photocurrent is achieved at light intensities as low as two equivalent suns. This strategy is simple, modular and offers unprecedented geometric and spatial control of the donor-acceptor interactions at an interface. These results are a key stepping stone toward the realization of an efficient TTA-UC solar cell that can circumvent the Shockley-Queisser limit.
Sigma-Aldrich
1,4-Dibromobenzene, 98%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Tin(II) oxide, ≤60 micron particle size, powder, 97%
Sigma-Aldrich
Anthraquinone, 97%
Sigma-Aldrich
Tin(II) oxide, 99.99% trace metals basis