Skip to Content
Merck
All Photos(3)

Key Documents

495425

Sigma-Aldrich

Gallium–Indium eutectic

Ga 75.5% / In 24.5%, ≥99.99% trace metals basis

Synonym(s):

EGaIn

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
11101711
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.99% trace metals basis

form

liquid

composition

Ga 75.5% / In 24.5%

mp

15.7 °C (lit.)

density

6.25 g/mL at 25 °C (lit.)

SMILES string

[Ga].[In]

InChI

1S/Ga.In

InChI key

SPAHBIMNXMGCMI-UHFFFAOYSA-N

General description

Gallium-indium eutectic 99.99% is a unique liquid metal alloy with a melting point of 15.7°C. It has a self-limiting oxide layer that retains its shape, making it ideal for use in various applications. This material is easy to mold, stretch, and form into a variety of shapes. It is also self-healing, which makes it useful in situations where the material may experience wear and tear. With its exceptional properties, gallium-indium eutectic 99.99% is a valuable material for a range of industries, including electronics, robotics, and medical devices.

Application

In the electronics industry, gallium-indium eutectic 99.99% is used as a thermal interface material due to its high thermal conductivity. It is also used as a flexible conductor in soft robotics and stretchable electronics. Because of its ability to conform to irregular shapes and high electronic conductivity, EGaIn is commonly used as an electrical contact for device testing and as the top contact in molecular electronics applications. Additionally, this liquid metal alloy is utilized in medical devices such as soft implants and wearable health monitors due to its biocompatibility and flexibility. Gallium-indium eutectic 99.99% has a wide range of potential applications, and its unique properties make it a valuable material for many industries.

Features and Benefits

  • Increase Power Performance: Its high thermal conductivity makes it a high-performing thermal interface material for increased power performance in electronics.

  • Simplify Electrical Contact: Its excellent electronic conductivity, conformal shape, and self-healing properties make it easy to make electrical contact for devices and molecular electronics

  • Robust Contact: This material conforms to irregular shapes, making it perfect for use in soft robotics and stretchable electronics as a flexible conductor and robust contact.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eutectic Gallium-Indium (EGaIn): A Liquid Metal Alloy for the Formation of Stable Structures in Microchannels at Room Temperature
Dickey MD, et al.
Advances in Functional Materials, 18(7), 1097-1104 null
Yu Long Han et al.
Scientific reports, 5, 11488-11488 (2015-07-02)
This paper describes a novel approach to fabricate paper-based electric circuits consisting of a paper matrix embedded with three-dimensional (3D) microchannels and liquid metal. Leveraging the high electric conductivity and good flowability of liquid metal, and metallophobic property of paper
On the use of Ga-In eutectic and halogen light source for testing P3HT-PCBM organic solar cells
Du Pasquier A, et al.
Solar Energy Mat. and Solar Cells, 90(12), 1828-1839 (2006)
Seoungwoong Park et al.
ACS nano, 14(7), 8485-8494 (2020-06-25)
Transition metal dichalcogenides (TMDs) have attracted significant interest as one of the key materials in future electronics such as logic devices, optoelectrical devices, and wearable electronics. However, a complicated synthesis method and multistep processes for device fabrication pose major hurdles
Nonreciprocal microwave devices based on magnetic nanowires.
Kuanr BK, et al.
Applied Physics Letters, 94(20) (2009)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the thermal conductivity of this alloy?

    1 answer
    1. The thermal conductivity of this eGaIn alloy - Ga 75.5% / In 24.5% - is cited in the literature as 26.43 W m−1 K−1.
      Please see the publication below for additional information:
      https://onlinelibrary.wiley.com/doi/10.1002/adma.202203391

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service