Skip to Content
Merck

263273

Gallium

99.999% trace metals basis

Synonym(s):

Elemental gallium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Ga
CAS Number:
Molecular Weight:
69.72
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141715
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

99.999% trace metals basis

form

(chips, chunks), (metallic liquid or solid (ambient temp dependent))

reaction suitability

core: gallium

resistivity

25.795 μΩ-cm, 30°C

bp

2403 °C (lit.)

mp

29.8 °C (lit.)

density

5.904 g/mL at 25 °C (lit.)

application(s)

semiconductor synthesis

storage temp.

2-8°C

SMILES string

[Ga]

InChI

1S/Ga

InChI key

GYHNNYVSQQEPJS-UHFFFAOYSA-N

Application

The wide liquid range of gallium was exploited recently when it was used as a solvent for the successful synthesis of ternary silicides, SmNiSi3 and YNiSi3, without incorporating Ga in their structure -- a promising new synthetic method.
Gallium (Ga) is a silver-colored metal that exists as a metallic liquid or solid, depending on the ambient temperature, with a melting point of 29.8 °C and a boiling point of 2,403 °C. This unique property allows gallium to be used in various applications, particularly in electronics and optoelectronics. Our high-purity gallium is characterized by its purity of 99.999% on a trace metals basis, it is utilized in the production of semiconductors, LED technology, and solar panels. Additionally, its exceptional thermal and electrical conductivity makes it particularly valuable in electronics and advanced technology applications.


Still not finding the right product?

Explore all of our products under Gallium


pictograms

CorrosionExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library