494143
Titanium(IV) diisopropoxidebis(2,2,6,6-tetramethyl-3,5-heptanedionate)
99.99%
Synonym(s):
Bis(dipivaloylmethanato)diisopropoxytitanium, Bis(isopropoxy)bis(dipivaloylmethanato)titanium, Diisopropoxybis(2,2,6,6-tetramethyl-3,5-heptanedionato)titanium, Titanium diisopropoxy bis(dipivaloylmethanate), Titanium(IV) diisopropoxide
About This Item
Recommended Products
Assay
99.99%
form
solid
reaction suitability
core: titanium
reagent type: catalyst
mp
171.1-184.6 °C (lit.)
SMILES string
CC(C)O[Ti](OC(C)C)(O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C
InChI
1S/2C11H20O2.2C3H7O.Ti/c2*1-10(2,3)8(12)7-9(13)11(4,5)6;2*1-3(2)4;/h2*7,12H,1-6H3;2*3H,1-2H3;/q;;2*-1;+4/p-2/b2*8-7-;;;
InChI key
FXUGBFBGCBGWKX-KKUWAICFSA-L
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.
Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.
Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.
Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service