Skip to Content
Merck
All Photos(3)

Key Documents

471038

Sigma-Aldrich

Lignosulfonic acid sodium salt

average Mw ~52,000, average Mn ~7,000

Synonym(s):

Sodium ligninsulfonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn ~7,000
average Mw ~52,000

composition

Na, 8 wt. %

impurities

4 wt. % reducing sugars
6 wt. %

solubility

H2O: soluble

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])CCCc1cc(c(cc1)OC(C[S](=O)(=O)O)Cc2c(c(ccc2)OC)[O-])OC

InChI

1S/C20H26O10S2.2Na/c1-28-18-7-3-6-15(20(18)21)12-16(13-32(25,26)27)30-17-9-8-14(11-19(17)29-2)5-4-10-31(22,23)24;;/h3,6-9,11,16,21H,4-5,10,12-13H2,1-2H3,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2

InChI key

YDEXUEFDPVHGHE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Dispersing agent for dyestuffs, wax emulsions, pigments, water treatment and cleaners.

Features and Benefits

Compatible with anionic and nonionic surfactants and wetting agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanyuan Ge et al.
International journal of biological macromolecules, 52, 300-304 (2012-10-31)
Different lignosulfonate (LS) samples were prepared via a three-step method and were characterized by FT-IR, UV, GPC and functional groups measurement. FT-IR and UV spectra confirmed the prepared samples had typical characteristics of lignin materials. GPC and functional groups measurement
Patricia Rodríguez-Lucena et al.
Journal of the science of food and agriculture, 91(3), 395-404 (2011-01-11)
During the last decade, environmental concerns regarding the use of recalcitrant synthetic chelates to overcome iron chlorosis has increased and new ligands such as lignosulfonates (LS) have been evaluated. However, the efficacy of these products is variable. In this work
Viorica Dulman et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 46(12), 1366-1375 (2011-09-29)
The potential of ammonium lignosulfonate (ALS) decolorization and degradation in aqueous solution was studied in a heterogeneous system using hydrogen peroxide and a Cu (II)-chelating ion exchanger. This was based on acrylic copolymers functionalized with N,N dimethylamino propylamine (DMAPA) as
Andrea Ertani et al.
Journal of agricultural and food chemistry, 59(22), 11940-11948 (2011-10-18)
Lignosulfonate-humate a and lignosulfonate-humate b, derived by an industrial process from lignin, were studied chemically and biologically, and their effects on maize metabolism compared with the responses induced by humic substances obtained from leonardite. Lignosulfonate-humate a and lignosulfonate-humate b elicited
Endry Nugroho Prasetyo et al.
Biotechnology journal, 7(2), 284-292 (2011-07-14)
This study combines the properties of siloxanes and lignin polymers to produce hybrid functional polymers that can be used as adhesives, coating materials, and/or multifunctionalized thin-coating films. Lignin-silica hybrid copolymers were synthesized by using a sol-gel process. Laccases from Trametes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service