69866
Montmorillonite
K 10, powder
Synonym(s):
K-Catalyst
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
powder
pH
3-4
SMILES string
[Si](=O)=O.[Si](=O)=O.[Si](=O)=O.[Si](=O)=O.[Al+3].[Al+3].[O-2].[O-2].[O-2].O
InChI
1S/2Al.4O2Si.H2O.3O/c;;4*1-3-2;;;;/h;;;;;;1H2;;;/q2*+3;;;;;;3*-2
InChI key
GUJOJGAPFQRJSV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Montmorillonite is a layered clay mineral (2:1 phyllosilicate) that belongs to a class of the smectite group. It is a naturally abundant material and is chemically modified by the cation-exchange method for the preparation of polymer nanocomposites.
Application
Montmorillonite has been used as:
It can also be used:
- A nanofiller to improve the mechanical properties of poly(vinyl alcohol) (PVA) film for potential usage in packaging applications.
- A catalyst in the conversion of polar and non-polar algae oil lipids to biodiesel.
It can also be used:
- As a nanofiller in the preparation of biopolymer based nanocomposites.
- In the preparation of Fe3+-montmorillonite, a solid acid catalyst for the synthesis of xanthenedione derivatives.
Other Notes
Solid Lewis acid catalyst
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Poly (vinyl alcohol): Montmorillonite: Boiled rice water (starch) blend film reinforced with silver nanoparticles; characterization and antibacterial properties.
Mathew S, et al.
Applied Clay Science, 161(1-2), 464-473 (2018)
O. Sieskind et al.
Tetrahedron Letters, 34, 1197-1197 (1993)
Thermal degradation kinetics and isoconversional analysis of biodegradable poly (3-hydroxybutyrate)/organomodified montmorillonite nanocomposites.
Achilias DS, et al.
Thermochimica Acta, 514(1-2), 58-66 (2011)
Biopolymer chitosan/montmorillonite nanocomposites: preparation and characterization.
Wang SF, et al.
Polymer Degradation and Stability, 90(1), 123-131 (2005)
Conversion of polar and non-polar algae oil lipids to fatty acid methyl esters with solid acid catalysts-A model compound study.
Asikainen M, et al.
Bioresource Technology, 191(1-2), 300-305 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service