470783
Vanadium(V) oxytripropoxide
98%
Synonym(s):
Vanadium(V) tripropoxide oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
OV(OC3H7)3
CAS Number:
Molecular Weight:
244.20
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
98%
form
liquid
reaction suitability
core: vanadium
reagent type: catalyst
bp
82-83 °C/0.25 mmHg (lit.)
density
1.076 g/mL at 25 °C (lit.)
SMILES string
CCCO[V](=O)(OCCC)OCCC
InChI
1S/3C3H7O.O.V/c3*1-2-3-4;;/h3*2-3H2,1H3;;/q3*-1;;+3
InChI key
QZCRZRGXAVPBNP-UHFFFAOYSA-N
Application
Vanadium (V) oxytripropoxide (VOTP) is often used as a precursor for V2O5. It may be used to prepare vanadia-silica mixed electrically conductive sol-gels for electrochemical regeneration of reduced disodium salt (NADH) for biocatalysis.
Analysis Note
Precursor for composite film electrodes.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
168.8 °F - closed cup
Flash Point(C)
76 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Electrochromic performance of sol?gel deposited V2O5:Ta films
Materials Science and Engineering, B, 138(2), 118-122 (2007)
Electrochemical Regeneration of NADH Using Conductive Vanadia-Silica Xerogels.
Siu E, et al.
Biotechnol. Progress, 23(1), 293-296 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service