Skip to Content
Merck
All Photos(1)

Key Documents

452130

Sigma-Aldrich

Poly(methyl methacrylate), isotactic

>80% isotactic

Synonym(s):

Isotactic PMMA, Isotactic methyl methacrylate polymer, Isotactic poly(methyl methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C5H8O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

density

1.2 g/mL at 25 °C (lit.)

SMILES string

[P](=O)(OCC)(OCC)OC(=O)\C(=N/OCC(=O)OC)\c1nc([s]c1)N

InChI

1S/C12H18N3O8PS/c1-4-21-24(18,22-5-2)23-11(17)10(8-7-25-12(13)14-8)15-20-6-9(16)19-3/h7H,4-6H2,1-3H3,(H2,13,14)/b15-10-

InChI key

DOXUHDDTLHKPOE-GDNBJRDFSA-N

General description

i-PMMA can be prepared from t-C4H9MgBr. Structure of crystalline isotactic poly(methyl methacrylate) (i-PMMA) was investigated by X-ray diffraction and infra red spectroscopic techniques.

Application

The polymeric blend of isotatic (i-) poly-(methyl methacrylate) & 6,13-bis(triisopropylsilylethynyl) pentacene can be used to prepare thin films transistors (TFTs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self?assembled organic channel/polymer dielectric layer for organic thin?film transistor applications.
Park JH, et al.
AIP Conference Proceedings, 847, 1399-1399 (2011)
Structure of isotactic poly (methyl methacrylate).
Tadokoro H, et al.
Macromolecules, 3(4), 441-447 (1970)
Kariem Mostafa Al Batouty et al.
The journal of adhesive dentistry, 15(2), 161-166 (2013-03-28)
To compare the push-out bond strength of a new glass fiber root canal filling material to that of Resilon/ Epiphany, EndoREZ, and gutta-percha/AH Plus root canal filling materials. Sixty roots were randomly divided into four groups (n = 15) according
S A Bini et al.
The bone & joint journal, 95-B(3), 367-370 (2013-03-02)
We evaluated the impact of pre-coating the tibial component with polymethylmethacrylate (PMMA) on implant survival in a cohort of 16 548 primary NexGen total knee replacements (TKRs) in 14 113 patients. In 13 835 TKRs a pre-coated tray was used
Panagiotis Korovessis et al.
Spine, 38(4), 292-299 (2013-02-15)
Prospective, parallel-group, controlled comparative randomized study. This study compares the efficacy in sagittal vertebral height and wedge deformity restoration, polymethylmethacrylate cement leakage safety, and functional outcome of balloon kyphoplasty (BK) versus KIVA (a novel vertebral augmentation technique) implant for the

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service