Merck
  • Push-out bond strength of a newly introduced glass fiber root canal filling material.
Push-out bond strength of a newly introduced glass fiber root canal filling material.
The journal of adhesive dentistry (2013-03-28)
Kariem Mostafa Al Batouty, Ahmed Abdel Hashem
ABSTRACT
To compare the push-out bond strength of a new glass fiber root canal filling material to that of Resilon/ Epiphany, EndoREZ, and gutta-percha/AH Plus root canal filling materials. Sixty roots were randomly divided into four groups (n = 15) according to the obturation material: group 1, Resilon/Epiphany (RE) (Pentron Clinical Technologies); group 2, gutta-percha/AH Plus (GP) (Dentsply DeTrey); group 3, EndoREZ obturation system (ER) (Ultradent); and group 4, glass fiber root canal filling (GF) (Stick Tech) and dual-curing resin sealer (MetaSeal, Parkell). Two-mm-thick horizontal sections from the coronal, middle, and apical thirds of each root were sliced for the push-out bond strength measurement. GF showed the significantly highest mean bond strength values at the coronal and middle thirds. At the apical third, GF showed significantly higher bond strength values than did RE and ER. However, there was no significant difference at the apical third between the GF and GP groups. The lowest bond strength values were recorded for ER. Glass fiber root canal filling material seems to provide promising bond strength to root canal dentin.
Product Description
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic