Skip to Content
Merck
All Photos(1)

Documents

439061

Sigma-Aldrich

(−)-Isopulegol

99%

Synonym(s):

(1R,2S,5R)-2-Isopropenyl-5-methylcyclohexanol, (1R,3R,4S)-p-Menth-8-en-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
1906573
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]20/D −22°, neat

refractive index

n20/D 1.471 (lit.)

bp

212 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1CC[C@H]([C@H](O)C1)C(C)=C

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h8-11H,1,4-6H2,2-3H3/t8-,9+,10-/m1/s1

InChI key

ZYTMANIQRDEHIO-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopulegol is monoterpene alcohol, a useful ingredient for the production of fragrances in perfume industries. It is also used as a starting material in the manufacture of menthol by the hydrogenation process. Menthol is an important component in cosmetics, pharmaceuticals, and toothpaste.

Application

(−)-Isopulegol can be used as a starting material for the enantioselective preparation of:      
  • 8-arylmenthols by Smiles-Truce rearrangement of aryl sulfonates.,·        
  • Stereoisomers of 5,9-dimethylpentadecane.
  • Octahydro-2H-chromen-4-ol by Prins cyclization with vanillin in the presence of montmorillonite clay as the catalyst.     
  • p-menthane-3,8,9-triol by catalytic Sharpless dihydroxylation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Green synthesis of (-)-isopulegol from (+)-citronellal: application to essential oil of citronella
Jacob RG, et al.
Tetrahedron Letters, 44(18), 3605-3608 (2003)
Enantioselective synthesis of three stereoisomers of 5, 9-dimethylpentadecane, sex pheromone component of Leucoptera coffeella, from (-)-isopulegol
Moreira JA and Correa AG
Tetrahedron Asymmetry, 14(23), 3787-3795 (2003)
Synthesis and Absolute Configuration at C (8) of `p-Menthane-3, 8, 9-triol?Derived from (-)-Isopulegol
Yuasa Y and Yuasa Y
Helvetica Chimica Acta, 87(10), 2602-2607 (2004)
Synthesis of octahydro-2H-chromen-4-ol from vanillin and isopulegol over acid modified montmorillonite clays: Effect of acidity on the Prins cyclization
Timofeeva MN, et al.
J. Mol. Catal. A: Chem., 398, 26-34 (2015)
Steven W M Crossley et al.
Organic letters, 18(11), 2620-2623 (2016-05-14)
Hydrogen atom transfer (HAT) circumvents a disfavored Friedel-Crafts reaction in the derivatization of the inexpensive monoterpene isopulegol. A variety of readily prepared aryl and heteroaryl sulfonates undergo a formal hydroarylation to form 8-arylmenthols, privileged scaffolds for asymmetric synthesis, as typified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service