Skip to Content
Merck
All Photos(2)

Key Documents

437905

Sigma-Aldrich

Dimethyl biphenyl-4,4′-dicarboxylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CC6H4C6H4CO2CH3
CAS Number:
Molecular Weight:
270.28
Beilstein:
2055852
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

213-215 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccc(cc1)-c2ccc(cc2)C(=O)OC

InChI

1S/C16H14O4/c1-19-15(17)13-7-3-11(4-8-13)12-5-9-14(10-6-12)16(18)20-2/h3-10H,1-2H3

InChI key

BKRIRZXWWALTPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl biphenyl-4,4′-dicarboxylate is a planar biphenyldicarboxylic acid ester. Crystal data of dimethyl biphenyl-4, 4′-dicarboxylate has been reported.

Application

Dimethyl biphenyl-4,4′-dicarboxylate may be employed as starting reagent for the synthesis of dimethyl 2-fluoro-and 2,2′-difluorobiphenyl-4,4′-dicarboxylates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimethyl biphenyl-4, 4'-dicarboxylate.
Ritzerfeld V, et al.
Acta Crystallographica Section E, Structure Reports Online, 65(7), 1677-1677 (2009)
Synthesis and properties of luminophores derived from fluorinated biphenyls.
Ol'khovik VK, et al.
Russ. J. Org. Chem., 44(8), 1172-1179 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service