Skip to Content
Merck
All Photos(1)

Key Documents

433071

Sigma-Aldrich

5-Chloro-1-indanone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7ClO
CAS Number:
Molecular Weight:
166.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

94-98 °C (lit.)

functional group

chloro
ketone

SMILES string

Clc1ccc2C(=O)CCc2c1

InChI

1S/C9H7ClO/c10-7-2-3-8-6(5-7)1-4-9(8)11/h2-3,5H,1,4H2

InChI key

MEDSHTHCZIOVPU-UHFFFAOYSA-N

General description

5-Chloro-1-indanone is a 5-halo-1-indanone. It participates in the Irie′s synthesis of substituted pyridines. 5-Chloro-1-indanone has a stable triclinic crystal structure and has intermolecular forces of C-H...O, C-H...Π, CO...Cl and Π...Π types.

Application

5-Chloro-1-indanone may be used as starting reagent for the preparation of 5-chloro-2-methoxycarbonyl-1-indanone. It may be used for the preparation of important biomedical compounds such as anticonvulsants, anticholinergics and diarylsulfonylureas, having potential activity against solid tumors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 5-Chloro-2-methoxycarbonyl-1-indanone [J].
HU X-G, et al.
Fine Chemicals / ????, 2, 022-022 (2009)
5, 6, 7-Trimethoxy-2, 3-dihydroinden-1-one.
Saeed A and Bolte M.
Acta Crystallographica Section E, Structure Reports Online, 63(5), 2757-2757 (2007)
Weak hydrogen-, halogen-and stacking Π....Π bonding in crystalline 5-chloro-1-indanone. An analysis by using X-ray diffraction, vibrational spectroscopy and theoretical methods.
Ruiz TP, et al.
Chemical Physics, 320(2), 164-180 (2006)
A facile synthesis of 7-halo-5H-indeno [1, 2-b] pyridines and-pyridin-5-ones.
DuPriest MT, et al.
The Journal of Organic Chemistry, 51(11), 2021-2023 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service