Skip to Content
Merck
All Photos(1)

Documents

11584

Sigma-Aldrich

(−)-Terpinen-4-ol

≥95.0% (sum of enantiomers, GC)

Synonym(s):

(R)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (R)-p-Menth-1-en-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D -25±6°, neat

optical purity

enantiomeric ratio: ~2:1

refractive index

n20/D 1.479

density

0.934 g/mL at 20 °C

SMILES string

CC(C)[C@@]1(O)CCC(C)=CC1

InChI

1S/C10H18O/c1-8(2)10(11)6-4-9(3)5-7-10/h4,8,11H,5-7H2,1-3H3/t10-/m0/s1

InChI key

WRYLYDPHFGVWKC-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terpinen-4-ol, a monoterpenoid alcohol, is the key constituent of tea tree (Melaleuca alternifolia) oil.

Application

(-)-Terpinen-4-ol has been used as a standard for the determination of the composition of Lavandula stoechas essential oil by fast gas chromatography-electronic impact ionization/mass spectrometry (FGC-EI/MS).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of the anticonvulsant activity of terpinen-4-ol
Sousa, Dami?o P. de, et al
Zeitschrift fur Naturforschung C, 64(1-2), 1-5 (2009)
Lavandula stoechas essential oil from Spain: Aromatic profile determined by gas chromatography-mass spectrometry, antioxidant and lipoxygenase inhibitory bioactivities.
Carrasco A, et al.
Industrial Crops and Products, 73, 16-27 (2015)
Zhenbiao Li et al.
Applied microbiology and biotechnology, 104(5), 2163-2178 (2020-01-26)
Tea tree oil (TTO) and its two characteristic components (terpinen-4-ol and 1,8-cineole) have been shown to inhibit Botrytis cinerea growth. In this study, we conducted a transcriptome analysis to determine the effects of TTO and its characteristic components, alone and
Laura de Fátima Souto Maior et al.
Clinical oral investigations, 23(7), 2837-2848 (2019-05-22)
This study investigated the anti-Candida activity and the Shore A hardness of a tissue conditioner (Softone™) modified by incorporation of terpinen-4-ol and cinnamaldehyde. Agar diffusion, microdilution, and mechanism of action methods were performed to determine to evaluate the antifungal activity
P H Hart et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 49(11), 619-626 (2000-12-29)
To evaluate potential antiinflammatory properties of tea tree oil, the essential oil steam distilled from the Australian native plant, Melaleuca alternifolia. The ability of tea tree oil to reduce the production in vitro of tumour necrosis factor-alpha (TNFalpha), interleukin (IL)-1beta

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service