Skip to Content
Merck
All Photos(2)

Key Documents

429252

Sigma-Aldrich

Poly(ethylene terephthalate)

granular

Synonym(s):

PET

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H8O4)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

abrasion resistance

30 mg (Taber, CS-17 wheel, 1kg, 50% RH, 1,000 cycles)

Quality Level

description

30% glass fiber to enhance hardness and strength

form

granular

hardness

120 (Rockwell R, ASTM D 785)

mp

250-255 °C

density

1.68 g/mL at 25 °C

SMILES string

OCCO.OC(=O)c1ccc(cc1)C(O)=O

InChI

1S/C8H6O4.C2H6O2/c9-7(10)5-1-2-6(4-3-5)8(11)12;3-1-2-4/h1-4H,(H,9,10)(H,11,12);3-4H,1-2H2

InChI key

FYIBGDKNYYMMAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in electrical parts including relay bases and lamp sockets, pump housings, gears, sprockets, chair arms, casters and furniture components.

Features and Benefits

Balance of strength, stiffness and toughness, excellent electrical properties, surface appearance and chemical resistance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark P Umstad et al.
The Australian & New Zealand journal of obstetrics & gynaecology, 50(5), 460-464 (2010-11-03)
Transabdominal cervical cerclage has been performed via laparotomy for over four decades. A laparoscopic approach has recently been developed and offers the potential for lower morbidity. The experience of one operator with transabdominal cervical cerclage via laparotomy is reviewed to
Simon Flury et al.
American journal of dentistry, 24(4), 226-232 (2011-10-25)
To investigate the surface hardness (Vickers hardness, HVN) of one light-curing flowable resin composite and five dual-curing resin cements after different polymerization procedures. The HVN was measured with a hardness-indentation device on one light-curing flowable resin composite (Tetric EvoFlow) and
Leonardo Pantoja Munoz et al.
Environmental science and pollution research international, 25(20), 20268-20279 (2018-06-28)
The introduction to the market of wet wipes, advertised and labelled as "flushable", has been the subject of controversy due to their perceived potential to block sewer systems as observed with other non-woven cloths such as traditional non-flushable wipes. Non-woven
Steven R Jefferies et al.
The Journal of clinical dentistry, 21(1), 20-23 (2010-06-10)
To evaluate the effect ofa nanofilled resin surface sealer (Seal&Protect) on the surface roughness of a hybrid composite resin. Twelve 15 mm x 2 mm disc composite resin specimens (TPH Spectrum Shade C-2) were fabricated and light-cured under controlled conditions.
Andrew J Stein et al.
Techniques in hand & upper extremity surgery, 15(1), 12-15 (2011-03-02)
Many surgical procedures have been described for the treatment of thumb basilar joint osteoarthritis. Augmentation of the standard ligament reconstruction tendon interposition procedure with the use of a Mersilene suture tape suspension-plasty, to recreate the stability provided by the anterior

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service