Skip to Content

1546900

USP

Polyethylene terephthalate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Poly(ethylene terephthalate)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C10H8O4)n
CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCCO.OC(=O)c1ccc(cc1)C(O)=O

InChI

1S/C8H6O4.C2H6O2/c9-7(10)5-1-2-6(4-3-5)8(11)12;3-1-2-4/h1-4H,(H,9,10)(H,11,12);3-4H,1-2H2

InChI key

FYIBGDKNYYMMAG-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Polyethylene terephthalate is the most commonly used polymer mainly in the manufacture of water bottles.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Used in electrical parts including relay bases and lamp sockets, pump housings, gears, sprockets, chair arms, casters and furniture components.

Features and Benefits

Balance of strength, stiffness and toughness, excellent electrical properties, surface appearance and chemical resistance.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Polyethylene terephthalate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library