Skip to Content
Merck
All Photos(1)

Documents

394505

Sigma-Aldrich

Xenon difluoride

99.99% trace metals basis

Synonym(s):

Xenon Fluoride (XeF2)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
XeF2
CAS Number:
Molecular Weight:
169.29
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

3.8 mmHg ( 25 °C)

Quality Level

Assay

99.99% trace metals basis

form

crystals

mp

129 °C (lit.)

density

4.32 g/mL at 25 °C (lit.)

SMILES string

F[Xe]F

InChI

1S/F2Xe/c1-3-2

InChI key

IGELFKKMDLGCJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xenon fluoride may be obtained by interacting elemental xenon and fluorine in the temperature range of 473-523 oC and 5 absolute atmosphere. Xenon difluoride readily interacts with Lewis acid and forms complexes.

Application

Very useful fluorination agent. Xenon fluoride may be used as a fluorinating agent to analyze sulphur, selenium and tellurium by gas chromatography.

Packaging

Packaged in PFA/FEP bottles

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorination with XeF(2).(1) 44. Effect of Geometry and Heteroatom on the Regioselectivity of Fluorine Introduction into an Aromatic Ring.
Marko Zupan et al.
The Journal of organic chemistry, 63(3), 878-880 (2001-10-24)
Infrared spectra of complex compounds of xenon difluoride with ruthenium pentafluoride
Prusakov VN, et al.
Journal of Applied Spectroscopy, 17(1), 920-922 (1972)
Use of Xenon difluoride for the determination of sulfur, selenium and tellurium as the volatile fluorides by gas chromatography
Aleinikov NN, et al.
Russian Chemical Bulletin, 22(11), 2552-2554 (1973)
Ariela W Kaspi et al.
Inorganic chemistry, 47(1), 5-7 (2007-12-07)
Palladium(II) aryliodo complexes bearing chelating diphosphine ligands react with XeF2, giving iodoarene and rare palladium(II) difluoro complexes. The reaction is general with regard to the aryl group, with even C6F5-I undergoing facile reductive elimination from a Pd center.
Minseob Kim et al.
Nature chemistry, 2(9), 784-788 (2010-08-24)
The application of pressure, internal or external, transforms molecular solids into extended solids with more itinerant electrons to soften repulsive interatomic interactions in a tight space. Examples include insulator-to-metal transitions in O(2), Xe and I(2), as well as molecular-to-non-molecular transitions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service