Skip to Content
Merck
All Photos(3)

Key Documents

372285

Sigma-Aldrich

2-Ethyl-9,10-dimethoxyanthracene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

117-119 °C (lit.)

SMILES string

CCc1ccc2c(OC)c3ccccc3c(OC)c2c1

InChI

1S/C18H18O2/c1-4-12-9-10-15-16(11-12)18(20-3)14-8-6-5-7-13(14)17(15)19-2/h5-11H,4H2,1-3H3

InChI key

SURWYRGVICLUBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethyl-9,10-dimethoxyanthracene (EDMA) has been reported as a potential anthracene photosensitizer for diaryliodonium and triarylsulfonium salts.
2-Ethyl-9,10-dimethoxyanthracene is reported to possess antihistaminic activity. Photoinduced electron transfer from the singlet excited state of 2-ethyl-9,10-dimethoxyanthracene to CCl4 has been reported. The photosensitized reductions of 4-cyanobenzylmethylphenyl sulfonium tetrafluoroborate by 2-ethyl-9,10-dimethoxyanthracene excited singlet state is reported to yield phenyl methyl sulfide.

Application

2-Ethyl-9,10-dimethoxyanthracene (EDA) has been used as sensitizer and its photoinduced (fluorescence quenching) reaction with 4-cyanobenzyl chloride has been studied.
2-Ethyl-9,10-dimethoxyanthracene may be employed as photosensitizer, to accelerate the photopolymerization. It may be employed as photosensitizer for enhancing the polymerization of benzothiazolium salts.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel N-methylbenzothiazolium salts as hardeners for epoxy and acrylate monomers.
Takahashi E, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 41(23), 3828-3837 (2003)
L Pause et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 1(4), 199-205 (2000-12-15)
It has been shown recently that the electrochemical reduction of carbon tetrachloride in N,N'-dimethylformamide follows a mechanism in which electron transfer and bond cleavage are concerted. We report here results concerning photoinduced electron transfer from the singlet excited state of
Photosensitized reduction of sulfonium salts: Evidence for nondissociative electron transfer.
Wang X, et al.
Journal of the American Chemical Society, 121(8), 4364-4368 (1999)
Stepwise and concerted pathways in photoinduced and thermal electron-transfer/bond-breaking reactions. Experimental illustration of similarities and contrasts.
Pause L, et al.
Journal of the American Chemical Society, 123(21), 4886-4895 (2001)
Anthracene electron-transfer photosensitizers for onium salt induced cationic photopolymerizations.
Crivello JV and Jang M.
Journal of Photochemistry and Photobiology A: Chemistry, 159(2), 173-188 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service