Recommended Products
Assay
98%
form
powder
mp
75-76 °C (lit.)
SMILES string
CC(=O)c1c2ccccc2cc3ccccc13
InChI
1S/C16H12O/c1-11(17)16-14-8-4-2-6-12(14)10-13-7-3-5-9-15(13)16/h2-10H,1H3
InChI key
NXXNVJDXUHMAHU-UHFFFAOYSA-N
Related Categories
Application
9-Acetylanthracene is used in the synthesis of luminescent moieties such as (9-anthryl)pyrazole (ANP) and 2,2-difluoro-4-(9-anthracyl)-6-methyl-1,3,2-dioxaborine. It is also used as a carrier ligand in the synthesis of fluorescent platinum(II) compounds.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Organic crystals with tunable emission colors based on a single organic molecule and different molecular packing structures.
Advanced Materials, 18(18), 2369-2372 (2006)
Luminescence and crystal structure of 2, 2-difluoro-4-(9-anthracyl)-6-methyl-1, 3, 2-dioxaborine.
Journal of Luminescence, 130(5), 756-761 (2010)
Synthesis, crystal structure, studies in solution and cytotoxicity of two new fluorescent platinum (II) compounds containing anthracene derivatives as a carrier ligand.
Inorganic Chemistry, 47(23), 11171-11179 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service