Skip to Content
Merck
All Photos(2)

Documents

370517

Sigma-Aldrich

1H,1H,2H,2H-Perfluorooctyl iodide

96%

Synonym(s):

1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl iodide, 8-Iodo-1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)5CH2CH2I
CAS Number:
Molecular Weight:
474.00
Beilstein:
1887105
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.359 (lit.)

bp

92 °C/45 mmHg (lit.)

density

1.934 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI

InChI

1S/C8H4F13I/c9-3(10,1-2-22)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h1-2H2

InChI key

NVVZEKTVIXIUKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>302.0 °F - closed cup

Flash Point(C)

> 150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel Rauber et al.
Physical chemistry chemical physics : PCCP, 19(40), 27251-27258 (2017-10-11)
Ionic liquids (ILs) exhibit tunable behaviour and properties that are due to their supramolecular structure. We synthesized a series of alkylated and fluorinated phosphonium dicyanamide ILs to study the relation between molecular structure and assembly with a focus on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service