Skip to Content
Merck
All Photos(1)

Documents

262544

Sigma-Aldrich

Heptadecafluoro-1-iodooctane

98%

Synonym(s):

1-Iodoperfluorooctane, Heptadecafluorooctyl iodide, Perfluorooctyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7I
CAS Number:
Molecular Weight:
545.96
Beilstein:
1717141
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

contains

copper pellets as stabilizer

refractive index

n20/D 1.329 (lit.)

bp

160-161 °C (lit.)

density

2.067 g/mL at 20 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

InChI

1S/C8F17I/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)26

InChI key

KWXGJTSJUKTDQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heptadecafluoro-1-iodooctane has been used as monodentate donor to investigate anion receptor complexes of mono-, bi- and tridentate donors with a variety of anions in the gas phase using both experimental and theoretical approaches.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth A L Gillis et al.
Physical chemistry chemical physics : PCCP, 15(20), 7638-7647 (2013-04-18)
Complexes of mono-, bi- (RB), and tridentate (RT) receptors with a range of anions (Cl(-), Br(-), I(-), NO3(-), H2PO4(-), HSO4(-), and tosylate (TsO(-))) have been studied in the gas phase by both experimental and theoretical methods. Temperature dependent blackbody infrared
Qian S Liu et al.
Environmental science & technology, 51(17), 10173-10183 (2017-07-27)
Per- and polyfluoroalkyl substances (PFASs) are a global concern because of their ubiquitous occurrence and high persistence in human blood, and increasing amounts of unidentified fluorinated compounds are now becoming new exposure issues. This study aims to investigate the structure-related
Wenting Song et al.
Talanta, 175, 413-420 (2017-08-27)
Contaminants of emerging concern are continuously increasing, which makes it important to develop high throughput screening techniques for the evaluation of their potential biological effects, especially endocrine disrupting effects, which would directly influence the population dynamics in environment. A novel
Ilona Bereczki et al.
ChemMedChem, 15(17), 1661-1671 (2020-07-12)
The limited scope of antiviral drugs and increasing problem of antiviral drug resistance represent a global health threat. Glycopeptide antibiotics and their lipophilic derivatives have emerged as relevant inhibitors of diverse viruses. Herein, we describe a new strategy for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service