Skip to Content
Merck
All Photos(3)

Key Documents

364304

Sigma-Aldrich

Nickel(II) chloride hydrate

99.95% trace metals basis

Synonym(s):

Nickel dichloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2 · xH2O
CAS Number:
Molecular Weight:
129.60 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.95% trace metals basis
form:
solid

vapor pressure

1 mmHg ( 671 °C)

Quality Level

Assay

99.95% trace metals basis

form

solid

composition

Degree of hydration, 7-8

reaction suitability

reagent type: catalyst
core: nickel

impurities

≤550.0 ppm Trace Metal Analysis

SMILES string

O.Cl[Ni]Cl

InChI

1S/2ClH.Ni.H2O/h2*1H;;1H2/q;;+2;/p-2

InChI key

HGGYAQHDNDUIIQ-UHFFFAOYSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengnan Wang et al.
Frontiers in chemistry, 1, 17-17 (2013-01-01)
The present research systematically investigated, for the first time, the transformation of sodium bicarbonate and CO2 into sodium formate over a series of Ni based metal nanoparticles (NPs). Ni NPs and eight NiM (M stands for a second metal) NPs
G D C Mendis et al.
Scientific reports, 9(1), 121-121 (2019-01-16)
Determining the mechanism of action (MOA) of novel or naturally occurring compounds mostly relies on assays tailored for individual target proteins. Here we explore an alternative approach based on pattern matching response profiles obtained using cultured neuronal networks. Conolidine and
Ritu Tyagi et al.
Journal of applied toxicology : JAT, 33(2), 134-141 (2011-09-20)
Heavy metals are known for their associated nephrotoxicity and nickel is no exception. An integrated metabonomic approach, based on high-resolution (1) H NMR spectroscopy, was applied to determine the acute biochemical effects of NiCl(2) on the renal tissues of rats.
Terran E Bergdale et al.
Journal of biotechnology, 161(3), 181-189 (2012-07-14)
Microbiologically induced calcium carbonate precipitation (MICCP) is a naturally occurring biological process that has shown its potential in remediation of a wide range of structural damages including concrete cracks. In this study, genetically engineered microorganisms, capable of producing extracellular polymeric
Steven J Ryan et al.
PloS one, 7(4), e35320-e35320 (2012-05-09)
The basolateral complex of the amygdala (BLA) is a critical component of the neural circuit regulating fear learning. During fear learning and recall, the amygdala and other brain regions, including the hippocampus and prefrontal cortex, exhibit phase-locked oscillations in the

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service