Skip to Content
Merck
All Photos(2)

Key Documents

203084

Sigma-Aldrich

Cobalt(II) chloride hydrate

99.999% trace metals basis

Synonym(s):

Cobalt(2+) chloride hydrate (1:2:1), Dichlorocobalt hydrate, cobalt dichloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2 · xH2O
CAS Number:
Molecular Weight:
129.84 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
powder and chunks
Pricing and availability is not currently available.

vapor pressure

40 mmHg ( 0 °C)

Quality Level

Assay

99.999% trace metals basis

form

powder and chunks

composition

Degree of hydration, 5-6

impurities

≤15.0 ppm Trace Metal Analysis

mp

86 °C (lit.)

density

1.924 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O.Cl[Co]Cl

InChI

1S/2ClH.Co.H2O/h2*1H;;1H2/q;;+2;/p-2

InChI key

QVCGXRQVUIKNGS-UHFFFAOYSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qilong Ren et al.
Nanoscale, 11(47), 23021-23026 (2019-11-28)
This article describes the fabrication of hollow manganese/cobalt oxide nanoparticles (MCO NPs) with a tunable size through a redox reaction and the Kirkendall effect for cancer imaging and drug delivery. MCO-70 NPs (with an average size of 70 nm) can
Geon-Young Yoo et al.
Analytical and bioanalytical chemistry, 412(12), 2873-2880 (2020-03-01)
Herein, we investigated the correlation between curcumin and glutathione (GSH) levels in mammalian cells using gold nanoparticles (AuNPs) and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS). GSH exists in high concentration in the cytosol and acts as a major

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service